Skip to Content
Merck
All Photos(1)

Documents

E1657

Sigma-Aldrich

EHT 1864

≥98% (HPLC)

Synonym(s):

5-(5-(7-(Trifluoromethyl)quinolin-4-ylthio)pentyloxy)-2-(morpholinomethyl)-4H-pyran-4-one dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H27F3N2O4S · 2HCl
CAS Number:
Molecular Weight:
581.47
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

H2O: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

Cl.Cl.FC(F)(F)c1ccc2c(SCCCCCOC3=COC(CN4CCOCC4)=CC3=O)ccnc2c1

InChI

1S/C25H27F3N2O4S.2ClH/c26-25(27,28)18-4-5-20-21(14-18)29-7-6-24(20)35-13-3-1-2-10-33-23-17-34-19(15-22(23)31)16-30-8-11-32-12-9-30;;/h4-7,14-15,17H,1-3,8-13,16H2;2*1H

InChI key

LSECOAJFCKFQJG-UHFFFAOYSA-N

Application

EHT 1864 has been used to study the regulation of the adapter protein dishevelled-1 ubiquitylation by Rac1 activity.

Biochem/physiol Actions

EHT 1864 induces dissociation of bound nucleotides. EHT 1864 induced inhibition of Rac prevents its association with downstream effectors. It is known to prevent estrogen-induced tumor cell proliferation in breast cancer.
EHT 1864 inhibits Rac function. It prevents Aβ40 and Aβ42 production in vitro without direct inhibition of g-secretase.

Features and Benefits

This compound is a featured product for Cyclic Nucleotide research. Click here to discover more featured Cyclic Nucleotide products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the GTP Binding Proteins (Low Molecular Weight) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Legal Information

EHT 1864 compound sold under license from ExonHit Therapeutics S.A. (www.exonhit.com)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Handbook of Cell Signaling, 2851-2851 (2009)
Jinkun Liu et al.
Evidence-based complementary and alternative medicine : eCAM, 2020, 2760979-2760979 (2020-05-19)
The infructescence of Platycarya strobilacea Sieb. et Zucc. (PS) has been used in the treatment of rhinitis and sinusitis in clinical practice. Our preliminary study showed that an ethanol extract of the infructescence of PS (EPS) had significant antinasopharyngeal carcinoma
Kara R Eichelberger et al.
mBio, 10(6) (2019-12-12)
Inhalation of Yersinia pestis causes primary pneumonic plague, the most severe manifestation of plague that is characterized by a dramatic neutrophil influx to the lungs. Neutrophils are ineffective during primary pneumonic plague, failing to control Y. pestis growth in the
Rac1 acts in conjunction with Nedd4 and dishevelled-1 to promote maturation of cell-cell contacts
Nethe M, et al.
Journal of Cell Science, jcs-100925 (2012)
Advances in Cancer Research, 134-134 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service