Skip to Content
Merck
All Photos(4)

Key Documents

DUO96010

Sigma-Aldrich

Duolink® PLA Multicolor Probemaker Kit - Red

Proximity Ligation Assay Multiplexing: protein-protein interaction, post-translational modification and low abundant protein events

Synonym(s):

in situ Proximity Ligation Assay Kit, Protein Protein Interaction Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105331
NACRES:
NA.32

product line

Duolink®

technique(s)

immunofluorescence: suitable
multiplexing: suitable
proximity ligation assay: suitable

fluorescence

λex 594 nm; λem 624 nm (Texas Red®)
λex 594 nm; λem 624 nm (red)

suitability

suitable for fluorescence

shipped in

dry ice

storage temp.

−20°C

Application

Specificity
Red fluorescence detection reagents are often used with Texas Red® filter.

Application Note
Primary antibodies are needed. Test your primary antibodies (IgG-class, mono- or polyclonal) in a standard immunofluorescence (IF), immunohistochemistry (IHC), or immunocytochemistry (ICC) assay to determine the optimal fixation, blocking, and titer conditions. Duolink® in situ reagents are suitable for use on fixed cells, cytospin cells, cells grown on slide, formalin-fixed, paraffin embedded (FFPE), or tissue (fresh or frozen). No minimum number of cells are required.

Let us do the work for you, learn more about our Custom Service Program to accelerate your Duolink® projects

View full Duolink® product list

Features and Benefits

Multicolor Duolink® proximity ligation assay (PLA®) is an application of the Duolink® technology that allows for endogenous, simultaneous detection of up to 4 protein interactions, post-translational modifications, or protein expression levels (including low abundant protein) in fixed cells and tissue samples. Follow the Duolink® PLA Probemaker Guide for Multicolor Detection and the Duolink® PLA Multicolor Detection protocol to use this product.

  • Visualize and detect multiple proteins events in one experiment.
  • No overexpression or genetic manipulation required
  • High specificity (fewer false positives)
  • Detection of low abundant proteins due to rolling circle amplification
  • Relative quantification possible
  • No special equipment needed
  • Faster and more simple than FRET
  • Increased accuracy compared to co-IP
  • Publication-ready results

Components
Duolink® PLA Multicolor Probemaker Kit - Red contains all the necessary reagents to perform the conjugations of Red Oligo A and Red Oligo B directly to primary antibodies. The resulting PLA probes (Red A and Red B) must be used together and cannot be paired with any other Duolink

This product is comprised of the following:
  • Red Oligo A DUO86010A
  • Red Oligo B DUO86010B
  • Conjugation Buffer DUO82033
  • Stop Reagent DUO82034
  • Storage Solution DUO82035

See datasheet for more information.

To perform a Multicolor Duolink® PLA experiment primary antibodies (PLA, IHC, ICC or IF validated) that recognize protein targets of interest are required. Duolink® PLA Multicolor Probemaker-generated probes (Red A/Red B, Green C/Green D, Orange F/Orange G, and/or FarRed H/FarRed I) are used in combination to perform the Multiplex experiment.The reagents in the Duolink® PLA Multicolor Reagent Pack are required to perform the amplification and detection of the resulting PLA probes. Required additional reagents include Duolink® Wash Buffers and Mounting Medium.

Legal Information

Duolink is a registered trademark of Merck KGaA, Darmstadt, Germany
PLA is a registered trademark of Merck KGaA, Darmstadt, Germany
Texas Red is a registered trademark of Life Technologies

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chatchai Phoomak et al.
Science advances, 7(3) (2021-02-02)
Asparagine (N)-linked glycosylation is required for endoplasmic reticulum (ER) homeostasis, but how this co- and posttranslational modification is maintained during ER stress is unknown. Here, we introduce a fluorescence-based strategy to detect aberrant N-glycosylation in individual cells and identify a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service