Skip to Content
Merck
All Photos(4)

Documents

DUO94105

Sigma-Aldrich

Duolink® flowPLA Mouse/Rabbit Starter Kit - Violet

Duolink® PLA kit for Flow Cytometry with Violet Detection and Mouse/Rabbit probes

Synonym(s):

Duolink Starter Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

Quality Level

technique(s)

flow cytometry: suitable
immunofluorescence: suitable
proximity ligation assay: suitable

fluorescence

λex 390 nm; λem 476 nm

suitability

suitable for fluorescence

storage temp.

−20°C

Specificity

Violet Fluorescence Detection Reagents
Use appropriate laser for λex 390 nm excitation
Use appropriate filter for λem 476 nm emission.

Application Note

Primary antibodies are needed. Test your primary antibodies (IgG-class, mono- or polyclonal) in a standard immunofluorescence (IF), immunohistochemistry (IHC), or immunocytochemistry (ICC) assay to determine the optimal fixation, blocking, and titer conditions. Flow validated antibodies are recommended.

Let us do the work for you, learn more about our Custom Service Program to accelerate your Duolink® projects.

View full Duolink® product list

Application

Based on proximity ligation assay (PLA), the Duolink® PLA technology allows for endogenous detection of protein interactions, post-translational modifications, and protein expression levels at the single molecule level in fixed cells.

Duolink® flowPLA Starter Kits will enable sensitive detection of proteins, protein-protein interactions, and protein modifications within cell populations by flow cytometry. To perform a Duolink® flowPLA experiment, you will need fixed, suspended cells, two primary antibodies that specifically recognize your proteins of interest and the Duolink® flowPLA starter Kit. The flowPLA Starter Kits are available with 3 different fluorophores: Violet, Green, and FarRed. The flowPLA starter Kits contain all the necessary reagents reagents for 100 Duolink® PLA reactions, including a pair of PLA probes (Anti-Rabbit PLUS and Anti-Mouse MINUS) and all the reagents to perform the amplification and detection of bound PLA probes by flow cytometry. Analysis is carried out using standard flow cytometry assay equipment. User must provide a fixed cell suspension, (rabbit and mouse-raised) primary antibodies, and corresponding PLA Probes.

Follow the Duolink® PLA Flow Cytometry Protocol to use this product.

Visit our Duolink® PLA Flow Cytometry page on how to run a Duolink® flow experiment, applications, troubleshooting, and more.

Features and Benefits

  • Analyze protein protein interactions with flow cytometry readout
  • Analyze cell populations with Proximity Ligation Assay
  • Increased sensitivity due to rolling circle amplification for low abundant targets
  • No overexpression or genetic manipulation required
  • Relative quantification possible
  • Works with any flow cytometer instrumentation
  • Easy to follow flexible protocol
  • Publication-ready results

Components

Duolink® flowPLA Detection Kit - Violet DUO94005-40TST
Duolink® In Situ PLA® Probe Anti-Mouse MINUS DUO92004-40TST
Duolink® In Situ PLA® Probe Anti-Rabbit PLUS DUO92002-40TST
Duolink® In Situ Wash Buffer, Brightfield (DUO82047-4L)

See datasheet for more information.

Legal Information

Duolink is a registered trademark of Sigma-Aldrich Co. LLC
PLA is a registered trademark of Sigma-Aldrich Co. LLC
Duolink is a registered trademark of Merck KGaA, Darmstadt, Germany
PLA is a registered trademark of Merck KGaA, Darmstadt, Germany
Plexiglas is a registered trademark of Arkema

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service