Skip to Content
Merck
All Photos(3)

Key Documents

DMN10

Sigma-Aldrich

GenElute Direct mRNA Miniprep Kits

sufficient for 10 purifications

Synonym(s):

GenElute Direct mRNA Miniprep Kit, GenElute mRNA Kit, Gen Elute

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105501
NACRES:
NA.52

usage

sufficient for 10 purifications

technique(s)

RNA purification: suitable

storage temp.

15-25°C

Looking for similar products? Visit Product Comparison Guide

General description

Procedures such as cDNA synthesis, expression profiling and others require separation of mRNA from the vastly more abundant rRNA and tRNA. The GenElute mRNA kits provide convenient procedures for isolating polyadenylated mRNA from previously prepared total RNA or directly from mammalian cells and tissues.

For direct mRNA preparation, cells or tissues are disrupted with SDS/proteinase K digestion to release RNA and eliminate RNases. The kit uses oligo (dT) covalently linked to 1 μm polystyrene beads to capture polyadenylated mRNA by hybridization. The polystyrene beads remain suspended during hybridization, eliminating the need for mixing or rocking, as is common for cellulose or magnetic particles. Polystyrene was also chosen because oligo(dT) polystyrene beads yield cleaner mRNA with fewer stringent washing steps than does the more commonly used oligo(dT) cellulose (2 or 3 wash steps versus 10 or more). With the GenElute kits, mRNA-bead complexes are washed on a microcentrifuge spin filter, and eluted into 10 mM Tris-HCL, pH 7.5.

Up to 107 mammalian cells or 40 mg tissue are lysed and homogenized, either with the filtration columns provided or with a mechanical homogenizer. RNase is eliminated during a 10 minute proteinase K digestion. Sodium chloride is added, and polyadenylated RNA is captured on oligo(dT) polystyrene beads during a 10 minute incubation. For further enrichment, RNA may be released from the beads into fresh lysis solution and recaptured with the original beads. After 3 washes in a spin column, purified mRNA is eluted in 100 μL of 10 mM Tris-HCl, pH 7.4.

The purified mRNA is ready for Northern analysis, reverse transcription and PCR, labeling for arrays, and other common applications.
The GenElute Direct mRNA Miniprep kit provides a convenient format to isolate polyadenylated mRNA directly from mammalian cells and tissues. The direct mRNA isolation procedure is based on that of Badley. Up to 107 mammalian cells or 40 mg tissue are lysed and homogenized, either with the filtration columns provided or with
a mechanical homogenizer. RNase is eliminated during a 10 minute proteinase K digestion. Sodium chloride is added, and polyadenylated RNA is captured on oligo(dT) polystyrene beads during a 10 minute incubation. For further enrichment, RNA may be released from the beads into fresh lysis solution and recaptured with the original beads. After 3 washes in a spin column, purified mRNA is eluted in 100 μl of 10 mM Tris-HCl, pH 7.4.

Application

The GenElute Direct Kit mRNA kit provides convenient procedures for isolating polyadenylated mRNA from previously prepared total RNA or directly from mammalian cells and tissues.

Features and Benefits

  • Poly (A)+ mRNA isolated from total RNA in 40 minutes or 60 minutes directly from cells and tissues
  • Oligo(dT) polystyrene beads require fewer wash steps
  • mRNA captured on oligo(dT) polystyrene beads in 10 minutes, with no mixing or rocking
  • mRNA captured on oligo(dT) polystyrene beads in 10 minutes, with no mixing or rocking (Fig. 1)
  • Poly (A)+ mRNA isolated from total RNA in 40 minutes (Fig. 2) or 60 minutes directly from cells and tissues (Fig. 3)
  • Oligo(dT) polystyrene beads require fewer wash steps

Other Notes

Use for isolating mRNA directly from mammalian cells or tissues.
For additional information, please see www.sigma-aldrich.com/mrna.

Legal Information

GenElute is a trademark of Sigma-Aldrich Co. LLC

Kit Components Only

Product No.
Description

  • Elution solution 1.5 mL

  • Filtration columns with tubes 10 ea

  • Lysis solution 20 mL

  • 5 M NaCl 1.5 mL

  • Oligo(dT)-polystyrene beads .3 mL

  • Proteinase K 5 mg

  • 40% Glycerol solution .6 mL

  • Spin columns with tubes 10 ea

  • Collection tube 10 ea

See All (9)

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Brahmchetna Singh et al.
Molecular cancer, 6, 82-82 (2007-12-26)
Retinoids are potent growth inhibitory and differentiating agents in a variety of cancer cell types. We have shown that retinoids induce growth arrest in all pancreatic cancer cell lines studied, regardless of their p53 and differentiation status. However, the mechanism
Abhirami Visvanathan et al.
Genes, 10(2) (2019-02-20)
Despite recent advances in N⁶-methyladenosine (m⁶A) biology, the regulation of crucial RNA processing steps by the RNA methyltransferase-like 3 (METTL3) in glioma stem-like cells (GSCs) remains obscure. An integrated analysis of m⁶A-RIP (RNA immunoprecipitation) and total RNA-Seq of METTL3-silenced GSCs

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service