Skip to Content
Merck
All Photos(2)

Key Documents

D9693

Sigma-Aldrich

Anti-Doublecortin antibody produced in rabbit

enhanced validation

~1 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-DCX, Anti-Doublecortex, Anti-LISX, Anti-X-linked lissencephaly, Anti-XLIS

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~40 kDa

species reactivity

human, mouse, rat

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

concentration

~1 mg/mL

technique(s)

western blot: 1-2 μg/mL using HEK-293T cells expressing human doublecortin. Also, mouse and rat brain extract (S1 fraction).

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DCX(1641)
mouse ... Dcx(13193)
rat ... Dcx(84394)

Immunogen

synthetic peptide corresponding to amino acids 151-170 of human doublecortin, conjugated to KLH. The corresponding sequence is identical in rat and mouse doublecortin.

Application

Anti-Doublecortin antibody produced in rabbit has been used in immunoblotting and immunohistofluorescence.
Anti-Doublecortin antibody produced in rabbit is suitable for immunoblotting at a working concentration of 1-2μg/mL using HEK-293T cells expressing human doublecortin, and mouse brain and rat brain extract (S1 fraction).

Biochem/physiol Actions

Doublecortin is a 40kDa microtubule-associated protein (MAP) that is essential for migration of neurons during the development of the cerebral cortex. Mutations in the human DCX gene cause lissencephaly (smooth brain) or subcortical laminar heterotopia (SCLH), characterized by mental retardation and seizures. DCX interaction with MAPs stabilizes microtubules (MTs). The interaction with MTs is via a conserved N-terminal doublecortin (DC) domain. DCX can be regulated by phosphorylation at multiple sites by several kinases, including JNK, Cdk5, PKA, and MARK/PAR-1 family of protein kinases.

Physical form

Solution in 0.01 M phophate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zuzana Fremuntova et al.
The European journal of neuroscience, 60(4), 4437-4452 (2024-06-18)
Mouse neuronal CAD 5 cell line effectively propagates various strains of prions. Previously, we have shown that it can also be differentiated into the cells morphologically resembling neurons. Here, we demonstrate that CAD 5 cells chronically infected with prions undergo
D Horesh et al.
Human molecular genetics, 8(9), 1599-1610 (1999-08-11)
X-linked lissencephaly is a severe brain malformation affecting males. Recently it has been demonstrated that the doublecortin gene is implicated in this disorder. In order to study the function of Doublecortin, we analyzed the protein upon transfection of COS cells.
María Alejandra González-González et al.
Scientific reports, 7, 40768-40768 (2017-01-21)
The periventricular zone of cerebellum is a germinative niche during the embryonic development, nevertheless its structural organization and functional implications in adult have not been widely studied. Here we disclose the presence of two novel clusters of cells in that
Orly Reiner et al.
Cell cycle (Georgetown, Tex.), 3(6), 747-751 (2004-05-01)
The mammalian cortex is generally subdivided into six organized layers, which are formed during development in an organized fashion. This organized cortical layering is disrupted in case of mutations in the doublecortin (DCX) gene. DCX is a Microtubule Associated Protein
J G Gleeson et al.
Neuron, 23(2), 257-271 (1999-07-10)
Doublecortin (DCX) is required for normal migration of neurons into the cerebral cortex, since mutations in the human gene cause a disruption of cortical neuronal migration. To date, little is known about the distribution of DCX protein or its function.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service