Skip to Content
Merck
All Photos(2)

Key Documents

D8394

Sigma-Aldrich

1,2-Dioleoyl-rac-glycerol

≥97%

Synonym(s):

(9Z)-9-Octadecenoic acid 1,1′-[1-(hydroxymethyl)-1,2-ethanediyl] ester, (±)-1,2-Diolein, 1,2-Di(cis-9-octadecenoyl)-rac-glycerol, rac-1,2-Dioleoylglycerol, rac-Glycerol 1,2-dioleate, DG(18:1(9Z)/18:1(9Z)/0:0)[rac]

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C39H72O5
CAS Number:
Molecular Weight:
620.99
Beilstein:
1917687
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25
Pricing and availability is not currently available.

biological source

synthetic (organic)

Quality Level

Assay

≥97%

form

liquid

impurities

≤3% 1,3-isomer

functional group

ester

lipid type

neutral glycerides

shipped in

dry ice

storage temp.

−20°C

SMILES string

CCCCCCCC\C=C/CCCCCCCC(=O)OCC(CO)OC(=O)CCCCCCC\C=C/CCCCCCCC

InChI

1S/C39H72O5/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-38(41)43-36-37(35-40)44-39(42)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h17-20,37,40H,3-16,21-36H2,1-2H3/b19-17-,20-18-

InChI key

AFSHUZFNMVJNKX-CLFAGFIQSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Diacylglycerol acyltransferases from Vernonia and Stokesia prefer substrates with vernolic acid.: Studies enzyme preferences for 1,2-Dioleoyl-rac-glycerol in the biosynthesis of industrially important oils, offering pathways to enhance bio-oil production using specific enzymatic processes (Yu et al., 2006).

Caution

Some isomerization may occur in storage or in solution.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Valeria Scala et al.
Frontiers in microbiology, 9, 1839-1839 (2018-08-30)
Lipids, components of the plasma and intracellular membranes as well as of droplets, provide different biological functions related to energy, carbon storage, and stress responses. Bacterial species display diverse membrane composition that changes in response to the different environmental conditions.
Xiumei Wang et al.
Bioprocess and biosystems engineering, 44(3), 575-584 (2020-11-21)
This study reports the properties of immobilized MAS1-H108A lipase from marine Streptomyces sp. strain W007 on XAD1180 resin and its application in the synthesis of n-3 polyunsaturated fatty acids (PUFA)-rich triacylglycerols (TAG) for the first time. It was found that
Xiumei Wang et al.
Journal of oleo science, 70(2), 227-236 (2021-01-19)
n-3 polyunsaturated fatty acids (PUFA)-rich triacylglycerols (TAG) with many beneficial effects are still difficult to be synthesized efficiently and rapidly by current synthetic techniques. This study reports the fatty acid specificity of immobilized MAS1 lipase and its efficient synthesis of
Ryutaro Adachi et al.
SLAS discovery : advancing life sciences R & D, 22(4), 360-365 (2017-03-23)
Monoacylglycerol acyltransferase (MGAT) activity catalyzes the synthesis of diacylglycerol (DAG) from fatty acyl-CoA and monoacylglycerol as substrates. It is important for the resynthesis of triacylglycerol (TAG) in the intestine. In the present study, we developed a MGAT enzymatic assay of
Xiumei Wang et al.
Bioresource technology, 235, 18-24 (2017-03-30)
This study reported a novel immobilized MAS1 lipase from marine Streptomyces sp. strain W007 for synthesizing high-yield biodiesel from waste cooking oils (WCO) with one-step addition of methanol in a solvent-free system. Immobilized MAS1 lipase was selected for the transesterification

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service