Skip to Content
Merck
All Photos(4)

Documents

D2006

Sigma-Aldrich

Dextrin from corn

Type I, powder

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

biological source

corn

type

Type I

form

powder

impurities

≤5% Reducing sugar

color

light yellow

SMILES string

OC[C@H]1O[C@H](O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C6H12O6/c7-1-2-3(8)4(9)5(10)6(11)12-2/h2-11H,1H2/t2-,3-,4+,5-,6+/m1/s1

InChI key

WQZGKKKJIJFFOK-DVKNGEFBSA-N

Gene Information

human ... PYGM(5837)

Looking for similar products? Visit Product Comparison Guide

Application

Dextrins from corn starch are used in nutrition studies to understand the effects of fiber in diets on energy levels, intestinal flora, digestion and other physiological processes. Dextrin may be used or derivatized for use in areas such as the stimulation of immune response and colitis research.

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Novel Fibers Increase Bone Calcium Content and Strength beyond Efficiency of Large Intestine Fermentation.
Weaver CM, Martin BR, Story JA, et al.
Journal of Agricultural and Food Chemistry, doi: 10-doi: 10 (2010)
Mamdouh N Samy et al.
Molecules (Basel, Switzerland), 24(12) (2019-06-19)
The strain Streptomyces osmaniensis CA-244599 isolated from the Comoros islands was submitted to liquid-state fermentation coupled to in situ solid-phase extraction with amberlite XAD-16 resin. Elution of the trapped compounds on the resin beads by ethyl acetate afforded seven metabolites
Maria L Stewart et al.
Annals of nutrition & metabolism, 56(2), 91-98 (2010-01-22)
Average dietary fiber intake in the United States is roughly half of the recommended amount. As new dietary fiber products are introduced to increase fiber intake, it is critical to evaluate the physiological effects of such fibers. This study examined
Julie-Anne Nazare et al.
Journal of the American College of Nutrition, 30(1), 63-72 (2011-06-24)
The effects of a new resistant dextrin ingested at breakfast on day-long metabolic parameters and ghrelin profile at subsequent lunch were investigated. In this randomized, single-blinded, crossover study, 12 healthy men ingested a standardized breakfast with 50 g of NUTRIOSE
Annet Maathuis et al.
Journal of the American College of Nutrition, 28(6), 657-666 (2010-06-03)
To investigate the effect of 5 newly developed maize-based fibers on the activity and composition of the microbiota in the colon. The fibers tested were glucose-based and had variable structures, including 2 resistant starch preparations, soluble corn fiber, pullulan, and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service