Skip to Content
Merck
All Photos(2)

Key Documents

D1194

Sigma-Aldrich

Anti-Derlin-2 antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-DERL2, Anti-Der1-like domain family, member 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~21 kDa

species reactivity

human, mouse

concentration

~1.0 mg/mL

technique(s)

indirect immunofluorescence: 5-10 μg/mL using HeLa cells
western blot: 2.0-4.0 μg/mL using whole extract of mouse 3T3 cells

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DERL2(51009)
mouse ... Derl2(116891)

Related Categories

General description

Derlin-1, Derlin-2, and Derlin-3 are the mammalian homologues of yeast Der1p, a transmembrane protein required for yeast endoplasmic reticulum-associated degradation (ERAD). Derlin-2 is approximately 30% identical to Derlin-1. In rat Derlin-2 is present to the endoplasmic reticulum(ER) membrane and forms a multisubunit complex with other proteins.

Immunogen

synthetic peptide corresponding to amino acid residues 223-239 of human derlin-2 conjugated to KLH via an N-terminal added cysteine residue. The corresponding sequence is identical in mouse.

Application

Anti-Derlin-2 antibody produced in rabbit has been used in immunoblotting and immunofluorescence.

Biochem/physiol Actions

Derlin-2, also known as F-LANa, is involved in the degradation of misfolded glycoproteins in the ER. Derlin-2 shares ~30% sequence identity with Derlin-1 and spans the lipid bilayer of the ER four times, showing structural similarity to Derlin-1. It is a component of the mammalian ER-associated degradation (ERAD) mechanism and is upregulated by unfolded protein response (UPR). Overexpression of this gene leads to increase in degradation of misfolded glycoprotein, whereas its knockdown blocks degradation. Derlin-2 also interacts with the mammalian orthologs of the yeast Hrd1p/Hrd3p ubiquitin-ligase complex.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Podocytes exhibit a specialized protein quality control employing derlin-2 in kidney disease
Ren G, et al.
American Journal of Physiology: Renal Physiology, 314(3), F471-F482 (2017)
Guohui Ren et al.
American journal of physiology. Renal physiology, 314(3), F471-F482 (2017-11-24)
Podocytes are terminally differentiated cells of the kidney filtration barrier with a limited proliferative capacity and are the primary glomerular target for various sources of cellular stress. Accordingly, it is particularly important for podocytes to cope with stress efficiently to
Murine polyomavirus requires the endoplasmic reticulum protein Derlin-2 to initiate infection
Lilley BN, et al.
Journal of Virology, 80(17), 8739-8744 (2006)
Brendan N Lilley et al.
Proceedings of the National Academy of Sciences of the United States of America, 102(40), 14296-14301 (2005-09-28)
Polypeptides that fail to pass quality control in the endoplasmic reticulum (ER) are dislocated from the ER membrane to the cytosol where they are degraded by the proteasome. Derlin-1, a member of a family of proteins that bears homology to
Yukako Oda et al.
The Journal of cell biology, 172(3), 383-393 (2006-02-02)
Proteins that are unfolded or misfolded in the endoplasmic reticulum (ER) must be refolded or degraded to maintain the homeostasis of the ER. Components of both productive folding and ER-associated degradation (ERAD) mechanisms are known to be up-regulated by the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service