Skip to Content
Merck
All Photos(1)

Key Documents

D0817

Sigma-Aldrich

(−)-Deguelin

>98% (HPLC), powder

Synonym(s):

(7aS,13aS)-13,13a-Dihydro-9,10-dimethoxy-3,3-dimethyl-3H-bis[1]benzopyrano[3,4-b:6′,5′-e]pyran-7(7aH)-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H22O6
CAS Number:
Molecular Weight:
394.42
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

>98% (HPLC)

form

powder

optical activity

[α]/D -70 to -80°, c = 0.2 in methanol

color

white to beige

mp

85-87 °C (lit.)

solubility

DMSO: 10 mg/mL, clear

storage temp.

−20°C

SMILES string

COc1cc2OC[C@H]3Oc4c(ccc5OC(C)(C)C=Cc45)C(=O)[C@H]3c2cc1OC

InChI

1S/C23H22O6/c1-23(2)8-7-12-15(29-23)6-5-13-21(24)20-14-9-17(25-3)18(26-4)10-16(14)27-11-19(20)28-22(12)13/h5-10,19-20H,11H2,1-4H3/t19-,20+/m1/s1

InChI key

ORDAZKGHSNRHTD-UXHICEINSA-N

Looking for similar products? Visit Product Comparison Guide

Application

(−)-Deguelin has been used as:
  • a nicotinamide adenine dinucleotide hydrogen (NADH) dehydrogenase (Complex I) inhibitor to study its effects on mitochondrial respiratory inhibition in human hepatocarcinoma cells (HepG2) and human renal proximal tubule epithelial cells (RPTECs)
  • an antiviral compound to study inhibitory effects on human cytomegalovirus (HCMV)-infected human foreskin fibroblast (HFF) cells
  • an analytical standard in high-performance liquid chromatography (HPLC)

Biochem/physiol Actions

Deguelin is a natural rotenoid compound present abundantly in barks, leaves, seeds, and roots of the plants belonging to the Leguminosae family. Deguelin has been studied as a therapeutic agent against the skin, lung cancer, and mammary tumorigenesis.
Inhibitor of activated Akt. Does not affect MAPK, ERK1/2, or JNK. Anticancer, chemoprotective agent.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhao-hui Chu et al.
Zhong xi yi jie he xue bao = Journal of Chinese integrative medicine, 9(5), 533-538 (2011-05-14)
To study the effects of deguelin on proliferation and apoptosis of human breast cancer cell line MCF-7 and on phosphatidylinositol 3-kinase/protein kinase B (PI3K/Akt) signaling pathway. After treatment with 0, 1, 5, 10, 15 and 20 μmol/L of deguelin for
Dong-Jo Chang et al.
Journal of medicinal chemistry, 55(24), 10863-10884 (2012-11-29)
Deguelin exhibits potent apoptotic and antiangiogenic activities in a variety of transformed cells and cancer cells. Deguelin also exhibits potent tumor suppressive effects in xenograft tumor models for many human cancers. Our initial studies confirmed that deguelin disrupts ATP binding
Rahel Sarah Varughese et al.
Cancer, 125(11), 1789-1798 (2019-04-02)
Deguelin is a rotenoid compound that exists in abundant quantities in the bark, roots, and leaves of the Leguminosae family of plants. An analysis of evidence from both in vitro and in vivo studies suggests that deguelin displays potent anticancer
Sebastian J Buss et al.
American journal of physiology. Heart and circulatory physiology, 302(2), H420-H430 (2011-11-08)
The attenuation of adverse myocardial remodeling and pathological left ventricular (LV) hypertrophy is one of the hallmarks for improving the prognosis after myocardial infarction (MI). The protein kinase Akt plays a central role in regulating cardiac hypertrophy, but the in
José Garcia et al.
Bioorganic & medicinal chemistry, 20(2), 672-680 (2011-11-01)
Deguelin, a rotenoid, has emerged as an attractive pharmacophore for chemoprevention showing in vivo activity in several xenografts. Recently, several lines of evidence have suggested its mode of action may involve inhibition of HSP90, however binding in a different mode

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service