This kit has only been evaluated with PC-PLC from mammalian sources.
CS0011
Phospholipase C (PLC) Activity Assay Kit
Sufficient for 100 colorimetric tests
Synonym(s):
Phospholipase C assay
About This Item
Recommended Products
General description
PC-PLC has a wide range of biological functions and is involved in various cell signalling pathways, including apoptosis, proliferation, differentiation, motility, gene expression and metastasis. Furthermore, it has been demonstrated that PC-PLC plays a crucial role in the development of atherosclerosis, making PC-PLC an interesting target for the development of cardiovascular drugs. Therefore, it is desirable to identify novel inhibitors of PC-PLC and to analyze their influence on PC-PLC.
Phospholipase C (PLC) Activity Assay Kit provides a simple, rapid and sensitive procedure for measuring PLC activity in various sample types. The kit can detect as low as 0.03 mU/well of PLC activity. The kit can detect PC-PLC activity of purified enzymes and PC-PLC activity in cell extracts. The kit can be used in screening PC-PLC inhibitors.
p-Nitrophenylphosphorylcholine is a chromogenic substrate that is used in this kit to measure the activity of PLC. Hydrolysis of p-nitrophenylphosphorylcholine by PLC results in the liberation of p-nitrophenol, which can be measured at 405 nm at pH 7.2-7.5. The colorimetric signal is proportional to the PLC activity.
Supportive calculator (Click here to download a calculator excel file): Will calculate the reagents needed according to your experimental needs.
Storage Class Code
10 - Combustible liquids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
Can this kit be used to detect the phospholipase activity of certain microorganisms?
1 answer-
Helpful?
-
-
Can this kit be used to measure PLC activity in human whole blood?
1 answer-
This kit is tested using cell lysates. It has not been tested on whole blood, however the dark color of whole blood would very likely mask results. It may be suitable for use with serum or plasma. The minimum limit of detection is 0.03 mU. It would be up to the end user to determine suitability.
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service