Skip to Content
Merck
All Photos(1)

Key Documents

CLS4594

Corning® 96 well plates

black/clear bottom, Fibronectin, lid, non-sterile

Synonym(s):

cell culture plate, multi well plates, multiple well plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
material:
black/clear bottom
size:
96 wells
Pricing and availability is not currently available.

material

black/clear bottom

sterility

non-sterile

feature

lid
Fibronectin

manufacturer/tradename

Corning 4594

size

96 wells

suitability

suitable for (cell cuture)

Looking for similar products? Visit Product Comparison Guide

Application

These plates are general assay plates, ideal for solution-based assays (i.e. hemagglutination or precipitation assays), serial dilutions and general storage applications.

Features and Benefits

  • Bottoms are 60% thinner than conventional polystyrene microplates, for sharp, clear images and minimal background in fluorescent assays
  • Opaque walls of microplates minimize well-to-well cross-talk
  • Optically clear flat bottom for direct microscopic viewing

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service