CLS4516
Corning® spheroid microplates
384 well, round black/clear bottom, ULA (Ultra-Low Attachment surface), lid, sterile
Synonym(s):
384 multiwell plate, 384 well microplate, 384 well microtiter plate, 384 well plate, EIA/RIA plate, immunoassay plate
About This Item
polystyrene
round black/clear bottom
Recommended Products
material
clear bottom
polystyrene
round black/clear bottom
sterility
sterile
feature
lid
skirt
Specific plates for 3D cell culture
ULA (Ultra-Low Attachment surface)
packaging
pkg of 5 ea
manufacturer/tradename
Corning 4516
size
384 wells
well volume
90 μL
wells
384 wells
working volume
20-80 μL
suitability
suitable for (cell cuture)
binding type
(ultra-low attachment)
Related Categories
General description
- Optically clear round bottom with black opaque microplate body
- Covalent attachment of Ultra-Low Attachment surface to reduce cellular adhesion to well surface
- Novel proprietary well geometry aids formation of spheroids in center of well
- Unique design that shields each well to minimize well-to-well cross talk
Key Benefits:
- Uniform, single spheroid formation across all wells
- Centering of the spheroid in each well enables automated visualization
- Culture and assay spheroids in the same microplate, without the need for transfer to a new plate
- Ease of media exchange or treatment with test compounds/drugs
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)