Skip to Content
Merck
All Photos(1)

Key Documents

CLS3913

Corning® Medium Bind 1 x 8 Stripwell 96 well plates

96 well plate, 1 x 8 Stripwell 8-well strips, Medium Bind, polystyrene, flat bottom, white, non-sterile, 100/cs

Synonym(s):

1 x 8 strip plate, 8 well strip plate, 96 multiwell plate, 96 well microplate, 96 well microtiter plate, 96 well plate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.15

material

flat bottom white wells
polystyrene plate
white polystyrene plate

sterility

non-sterile

feature

lid: no
skirt
plate format: 96 well Stripwell 8-well strips

packaging

case of 100
pack of 25

manufacturer/tradename

Corning 3913

size

96 wells

well volume

360 μL

well working volume

75-200 μL

color

white

suitability

suitable for (in vitro diagnostic assays)

binding type

non-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

Corning® Medium Bind 1 x 8 Stripwell 96 Well Plates

  • Designed for in vitro diagnostic assays
  • Flat bottom strips are designed to easily break apart and are pre-assembled in an "egg-crate" style strip holder that allows each individual well to be positioned back into the plate once broken
  • Black wells have low background fluorescence, minimal light scatter and reduced crosstalk
  • White wells enhance luminescent signals, have low background luminescence and fluorescence, and prevent well-to-well crosstalk
  • Not treated (or medium binding) polystyrene surface is hydrophobic in nature and binds biomolecules through passive interactions
  • Is suitable primarily for the immobilization of large molecules, such as antibodies, that have large hydrophobic regions that can interact with the surface
  • Untreated microplates have a binding capacity of approximately 100 to 200ng IgG/cm2
  • Recommended working volume of 75 to 200 μL
  • 1x8 Strips fit only one way into the strip holder, eliminating the chance of misorientation

Legal Information

Corning is a registered trademark of Corning, Inc.
Stripwell is a trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service