CLS3643
Corning® 384 well microplate, low volume
poly-D-lysine coated surface, black polystyrene, round bottom, sterile, lid, pack of 10
Synonym(s):
384 multiwell plates, 384 well low volume plates, 384 well microplates, 384 well microtiter plates, 384 well plates, 384 well tissue culture treated plates
About This Item
flat bottom clear polystyrene wells
round bottom
Recommended Products
material
black polystyrene
flat bottom clear polystyrene wells
round bottom
sterility
sterile
feature
lid
skirt
packaging
pack of 10
case of 50
manufacturer/tradename
Corning 3643
size
384 wells
surface area
0.03 cm2 , cell growth area
well volume
50 μL
well working volume
5-40 μL
wells
384 wells , low volume
color
black
suitability
suitable for (molecular, validated for the FLIPR reader)
binding type
poly-D-lysine coated surface
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
- Round well design with a clear flat bottom
- 2.8 mm distance between well bottoms and plate bottom
- 50μL total well volume, with recommended working volume of 5-40μL
- High base plate design is compatible with automation and for bar coding
- Validated by Molecular Devices for the FLIPR reader. Outer wells aligned for proper viewing with FLIPR
- Poly-D-Lysine coated for enhanced cell attachment
- Sterile (aseptic assembly) and certified nonpyrogenic
- Ideal for miniaturized 384 well fluorescent cell-based assays
Note: Suitable for 384 well fluorescent cell-based assays
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service