Skip to Content
Merck
All Photos(1)

Documents

CLS354241

Corning® Cell-Tak(TM) Cell and Tissue Adhesive

pack of 5 mg

Synonym(s):

Tissue Adhesive

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116155
NACRES:
NA.7

packaging

pack of 5 mg

manufacturer/tradename

Corning 354241

Looking for similar products? Visit Product Comparison Guide

General description

Corning Cell-Tak Cell and Tissue Adhesive, 5 mg, is used to attach cells or tissue sections to many types of surfaces, including plastic, glass, metal, FEP Polymer, and biological materials.
Quality
Dopa: Protein ratio > 0.05

Source
Polyphenolic proteins secreted by Mytilus edulis

Molecular Weight
110-140 kD.

Preparation and Storage
Stable for at least three months at 2° to 8°C. Do not freeze.

Handling
Recommended concentration - 1-5 μg/cm² of growth surface depending on cell type (guidelines for coating included with product).

Formulation
As a liquid in 5% acetic acid

Legal Information

Corning is a registered trademark of Corning, Inc.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service