CLS3526
Corning® Costar® TC-Treated Multiple Well Plates
size 24 wells, polystyrene plate, flat bottom wells, case of 50 (individually wrapped), sterile, lid
Synonym(s):
cell culture plate, multi well plates, multiple well plates, multiwell plates, tissue culture plates
About This Item
flat bottom wells
polystyrene plate
round clear wells
skirt
Recommended Products
material
clear bottom
flat bottom wells
polystyrene plate
round clear wells
description
growth area 1.9 cm2
sterility
sterile
feature
lid
skirt
packaging
pack of 1 (individually wrapped)
case of 50 (individually wrapped)
manufacturer/tradename
Corning 3526
size
24 wells
well diam.
15.6 mm
well volume
3.4 mL
well working volume
0.38-0.57 mL
binding type
Tissue Culture (TC)-treated surface
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service