CLS352027
Corning® Falcon® Round Bottom Test Tubes
capacity 8 mL, cap, screw, sterile
Synonym(s):
culture tube
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41121700
NACRES:
NB.13
Recommended Products
material
round polystyrene
sterility
sterile
feature
cap screw
manufacturer/tradename
Corning 352027
capacity
8 mL
Looking for similar products? Visit Product Comparison Guide
Application
Liquid Handling
Legal Information
Corning is a registered trademark of Corning, Inc.
Falcon is a registered trademark of Corning, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service