Skip to Content
Merck
All Photos(2)

Key Documents

CLS3362

Corning® 96 Well Solid Polystyrene Microplate

flat bottom, white polystyrene, bag of 25, sterile, lid: no

Synonym(s):

96 multiwell plates, 96 well microplates, 96 well microtiter plates, 96 well plates

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.13

material

flat bottom
white bottom
white polystyrene

sterility

sterile

feature

lid: no

packaging

bag of 25

manufacturer/tradename

Corning 3362

size

96 wells

surface area

0.32 cm2 , cell growth area

cell growth area

cell growth area

well volume

360 μL

working volume

75-200 μL

color

white

binding type

Tissue Culture (TC)-treated surface

Looking for similar products? Visit Product Comparison Guide

General description

  • Corning® 96 Well Solid Polystyrene Microplate White walled plates with no lid have flat bottoms with a total volume of 360 μL.
  • Enhances luminescent signals.
  • Displays low background luminescence and fluorescence.
  • Recommended working volumes of 75 to 200 μL.
  • Treated for optimal cell attachment
  • Nonpyrogenic and gamma radiation sterilized.
  • Individual alphanumeric codes enable well identification.

Legal Information

Corning is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service