CLL1158
A549 Cells GFP-STAT1
human male lung (Source Disease: Lung carcinoma)
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
UNSPSC Code:
41106514
NACRES:
NA.81
Recommended Products
Product Name
A549 Cells GFP-STAT1,
biological source
human male lung (Source Disease: Lung carcinoma)
OMIM accession no.
storage temp.
−196°C
Gene Information
human ... STAT1(6772)
General description
A549 GFP-STAT1 are lung carcinoma cells from a human 58 year old caucasian male having two distinct ZFN modifications creating two fluorescent transgenes each expressed from their endogenous gene locus.
This cell line was derived from ATCC Catalog No. CCL-185.
This cell line was derived from ATCC Catalog No. CCL-185.
Application
This product is a human A549 cell line in which the genomic STAT1 gene has been endogenously tagged with a Green Fluorescent Protein (GFP) gene using CompoZr® zinc finger nuclease (ZFN) technology. Integration resulted in endogenous expression of the fusion protein in which GFP is attached to the C-terminus of STAT1. Fluorescence imaging shows STAT1-GFP expression throughout the cell. Upon activation with a ligand such as interferon-gamma (INF-γ), the cell line shows redistribution of STAT1 from the cytoplasm to the nucleus, making it useful for high content screening of compounds that modulate STAT1 activity. This stable cell line was expanded from a single clone. The target′s gene regulation and corresponding protein function are preserved in contrast to cell lines with overexpression via an exogenous promoter.
To learn more, please visit the Cellular Reporter Cell Line webpage
Features and Benefits
Zinc Finger Nuclease (ZFN)-mediated targeted integration of a fluorescent GFP tag into the last coding exon just preceding the stop codon of the STAT1 gene on chromosome 2q32.2 to create a cell line exhibiting stable expression of the GFP transgene.
The A549 cells are adherent, with a doubling time of approx. 22 hours.
The A549 cells are adherent, with a doubling time of approx. 22 hours.
Quality
Tested for Mycoplasma, sterility, post-freeze viability, short terminal repeat (STR) analysis for cell line identification, cytochrome oxidase I (COI) analysis for cell line species confirmation.
Preparation Note
Media Renewal changes two to three times per week.
Rapidly thaw vial by gentle agitation in 37°C water bath (~2 minutes), keeping vial cap out of the water. Decontaminate with 70% ethanol, add 9 mL culture media and centrifuge 125 x g (5-7 minutes). Resuspend in complete culture media and incubate at 37°C in a 5% CO2 atmosphere.
Subculture Ratio: approx. 1:3-1:8
The base medium for this cell line is F12 Ham Medium, Cat. No. N4888. To make the complete growth medium, add the following components to the base medium: fetal bovine serum, Cat. No. F4135, to a final concentration (v/v) of 10% and L-glutamine, Cat. No. G7513, at a final concentration of 2 mM.
Cell freezing medium-DMSO 1X, Cat. No. C6164.
Rapidly thaw vial by gentle agitation in 37°C water bath (~2 minutes), keeping vial cap out of the water. Decontaminate with 70% ethanol, add 9 mL culture media and centrifuge 125 x g (5-7 minutes). Resuspend in complete culture media and incubate at 37°C in a 5% CO2 atmosphere.
Subculture Ratio: approx. 1:3-1:8
The base medium for this cell line is F12 Ham Medium, Cat. No. N4888. To make the complete growth medium, add the following components to the base medium: fetal bovine serum, Cat. No. F4135, to a final concentration (v/v) of 10% and L-glutamine, Cat. No. G7513, at a final concentration of 2 mM.
Cell freezing medium-DMSO 1X, Cat. No. C6164.
Legal Information
CompoZr is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service