Skip to Content
Merck
All Photos(1)

Key Documents

SSN-1

96082808, fish unknown/unspecified, Fibroblast

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
biological source:
fish unknown/unspecified
growth mode:
Adherent
karyotype:
Aneuploid
morphology:
Fibroblast
products:
Not specified
receptors:
Not specified

Product Name

SSN-1, 96082808

biological source

fish unknown/unspecified

growth mode

Adherent

karyotype

Aneuploid

morphology

Fibroblast

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Fish striped snakehead fry, spontaneous production of c-type retrovirus

Cell Line Description

The fish cell line SSN-1 was initiated from whole fry tissue, Channa (Ophicephalus) striatus, commonly named ′striped snakehead′. SSN-1 spontaneously produce and release endogenous snakehead fish Mn²+- dependent C-type retrovirus. The cell line is suitable for fish virus isolation and is susceptible to epizootic ulcerative syndrome rhabdoviruses and piscine nodaviruses. SSN-1 was previously infected with mycoplasma but has been eradicated and tested negative after a minimum of 10 passages.

Application

Isolation of fish virus pathogens

Culture Medium

L15 + 2mM Glutamine + 5-10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA. Use trypsin very cautiously; 25-30°C, can be light sensitive therefore avoid prolonged exposure to bright light. Fish cell lines detach easily during transit if the culture is very young. After resuscitation split cells 1:3 to 1:4 and culture for at least one week with 1-2 media changes before shipping.

When resuscitating a frozen ampoule of cells we recommend the cells are seeded at the higher seeding level i.e. 4 x10, 000/cm². Cells may take up to 10 days to reach 70% following resuscitation and subculture. Therefore media change flasks after 4-5 days to encourage growth.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service