AT-2.1
94101450, rat prostate (ventral), Epithelial-like
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106514
Recommended Products
Product Name
AT-2.1, 94101450
biological source
rat prostate (ventral)
packaging
tube of 5 μg 94101450-DNA-5UG
pkg of vial of cells 94101450-1VL
growth mode
Adherent
karyotype
Not specified
morphology
Epithelial-like
products
Not specified
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
cancer
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Rat prostatic cancer
Cell Line Description
AT-2.1 was developed via in vivo passages of the Dunning R-3327 rat prostatic cancers. A series of other cell lines were produced and they hold in common with Dunning R-3327 and AT-2.1 a structural change in chromosome 4 involving a translocation.
Culture Medium
RPMI 1640 + 2mM Glutamine + 250nM Dexamethasone (DXMT) + 10% Foetal Bovine Serum (FBS).
Subculture Routine
Split sub-confluent cultures (70-80%) 1:3 to 1:10 i.e. seeding at 1-4x10,000 cells/ml. Cells can be easily dislodged from the monolayer mechanically, however 0.25% trypsin or trypsin/EDTA can be used; 5% CO2; 37°C.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service