Skip to Content
Merck
All Photos(1)

Key Documents

P1.HTR

87121104, mouse breast, Mastocytoma

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

P1.HTR, 87121104, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

biological source

mouse breast

packaging

tube of 5 μg 87121104-DNA-5UG
pkg of vial of cells 87121104-1VL

growth mode

Suspension

karyotype

Not specified

morphology

Mastocytoma

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse DBA/2 mastocytoma

Cell Line Description

From P815 after serial cycles of transfection. This HTR variant shows high efficiency of transfection (0.00001) for neomycin-resistance and TK gene or other exogenous DNA. Possible to select TK- cells with BUdR for resubmission to transfection with TK gene.

Application

Genetic, transfection and tumourigenicity studies

Culture Medium

DMEM + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 8% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service