Skip to Content
Merck
All Photos(1)

Key Documents

Y3.AG.1.2.3

85110502, rat blood, Not specified

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

Y3.AG.1.2.3, 85110502

biological source

rat blood

growth mode

Suspension

karyotype

Not specified

morphology

Not specified

products

Immunoglobulin G (IgG) kappa light chains

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Rat LOU myeloma

Cell Line Description

Sub-cloned from an azaguanine-resistant mutant of the Lou rat myeloma, S210. The cells secrete IgG kappa light chains.

Application

Tumourigenicity studies

Culture Medium

RPMI 1640 + 2mM Glutamine + 1mM Sodium Pyruvate (NaP) + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service