Skip to Content
Merck
All Photos(1)

Documents

RN46A-B14

12061303, rat embryo d13 medullary raphe. Morphology: proliferating, fibroblast. Differentiating, neuronal.

Synonym(s):

46A B14, 46A-B14, RN46A B14

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

RN46A-B14, 12061303

biological source

rat embryo (day 13 medullary raphe)

growth mode

Adherent

karyotype

Not specified

morphology

Fibroblast morphology while proliferating and neuronal on differentiation

receptors

Not specified

Looking for similar products? Visit Product Comparison Guide

Cell Line Origin

Embryonic rat medullary raphe, temperature-sensitive mutant of SV40 large T-antigen, immortalised, serotonergic, neuronal

Cell Line Description

The clonal cell line, RN46A-B14, was isolated following transfection of the gene encoding rat brain-derived neurotrophic factor (BDNF) into RN46A cells (see ECACC Catalogue number 12061302). RN46A-B14 cells synthesize and secrete biologically active BDNF in vitro and synthesize serotonin (5-HT) following partial membrane depolarization. Two weeks following RN46A-B14 cell transplantation into the adult rat cortex and hippocampus, there is a threefold increase in survival of RN46A-B14 cells compared to RN46A cells. The grafted RN46A-B14 cells i mMunohistochemically stain for BDNF and 5-HT, while RN46A cells transfected with vector only are negative for both BDNF and 5HT. In addition, RN46A-B14 cells attain more morphologically complex phenotypes, indicating enhanced neuronal differentiation. Autocrine secretion of BDNF by RN46A-B14 cells thus potentiates survival and can be used to deliver both BDNF and 5-HT in vivo.

DNA Profile

Not specified

Culture Medium

DMEM:F12 (1:1) (D8062) + L-Glutamine (G7513) + 10% FBS / FCS (F2442) + 0.25mg/ml Geneticin (G418) + 0.1mg/ml hygromycin. Alternatively CNS medium can be used (see Kawamoto & Barrett 1986).

Subculture Routine

Split subconfluent cultures (70-80%) 1:2 to 1:5 using 0.25% trypsin/EDTA; 5% CO2; 33 °C. Suggested seeding density 2-4 x 10,000 cells/cm2. Doubling time 25hrs.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service