Skip to Content
Merck
All Photos(1)

Key Documents

LA1-55n

6041203, human nerve, Fusiform cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

LA1-55n, 06041203

biological source

human nerve

growth mode

Semi-adherent aggregates

karyotype

Not specified

morphology

Fusiform cells

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human Neuroblastoma

Cell Line Description

Derived from LA1-5s a clonal subline of LA-N-1. LA-N-1 was derived from neuroblastoma cells in the bone marrow of a 2-year-old male with clinical Stage IV neuroblastoma.LA-N-1 (Sigma Catalogue number 06041201), LA1-55n (Sigma Catalogue number 06041203) and LA1-5s (Sigma Catalogue number 06041204) have been shown to originate from the same patient by STR profiling.

Culture Medium

EMEM (with non-essential amino acids) and Ham′s F12 (1:1 mixture) + 2mM Glutamine + 10% Foetal Bovine Serum (FBS)

Subculture Routine

For routine maintenance, split cultures only after they have become very dense i.e. split once every 2-3 weeks at a 1:20 to 1:100 ratio; 8% CO2, 37°C. Cells are moderate to weakly adherent. Allow floating clumps to settle then withdraw medium. Remove at

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service