Skip to Content
Merck
All Photos(1)

Key Documents

C7515

Sigma-Aldrich

Cannabidiol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H30O2
CAS Number:
Molecular Weight:
314.46
MDL number:
UNSPSC Code:
12352200

form

powder

drug control

USDEA Schedule I; regulated under CDSA - not available from Sigma-Aldrich Canada

storage temp.

2-8°C

SMILES string

Oc1c(c(cc(c1)CCCCC)O)C2[C@@H](CCC(=C2)C)C(=C)C

InChI

1S/C21H30O2/c1-5-6-7-8-16-12-19(22)21(20(23)13-16)18-11-15(4)9-10-17(18)14(2)3/h11-13,17-18,22-23H,2,5-10H2,1,3-4H3/t17-,18?/m0/s1

InChI key

QHMBSVQNZZTUGM-ZENAZSQFSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Not psychoactive in humans.

Other Notes

Major component of marijuana from C. sativa

comparable product

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shaher Duchi et al.
Journal of neuroimmunology, 258(1-2), 32-40 (2013-03-23)
We investigated the efficiency of nasal drug administration as a new non-invasive treatment strategy for MS. Glatiramer Acetate (GA) and GA-Cannabidiol (CBD) combination administered in nasal delivery system (NDS) resulted in a statistically significant decrease of clinical scores and inflammatory
Alline Cristina Campos et al.
Psychopharmacology, 226(1), 13-24 (2012-09-26)
Cannabidiol (CBD) is a non-psychotomimetic constituent of Cannabis sativa plant that promotes antianxiety and anti-panic effects in animal models after acute systemic or intra-dorsal periaqueductal gray (DPAG) administration. However, the effects of CBD repeated administration, and the possible mechanisms involved
Sergiy Sylantyev et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(13), 5193-5198 (2013-03-09)
G protein-coupled receptor (GPR) 55 is sensitive to certain cannabinoids, it is expressed in the brain and, in cell cultures, it triggers mobilization of intracellular Ca(2+). However, the adaptive neurobiological significance of GPR55 remains unknown. Here, we use acute hippocampal
Tatiana Barichello et al.
European journal of pharmacology, 697(1-3), 158-164 (2012-10-23)
Pneumococcal meningitis is a life-threatening disease characterized by an acute infection affecting the pia matter, arachnoid and subarachnoid space. The intense inflammatory response is associated with a significant mortality rate and neurologic sequelae, such as, seizures, sensory-motor deficits and impairment
Stephen D Skaper et al.
Philosophical transactions of the Royal Society of London. Series B, Biological sciences, 367(1607), 3193-3200 (2012-10-31)
The psychoactive component of the cannabis resin and flowers, delta9-tetrahydrocannabinol (THC), was first isolated in 1964, and at least 70 other structurally related 'phytocannabinoid' compounds have since been identified. The serendipitous identification of a G-protein-coupled cannabinoid receptor at which THC

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service