C6039
Cell Freezing Medium-Glycerol 1×
sterile-filtered, suitable for cell culture
Synonym(s):
Cell Cryopreservation Solution, Freezing Medium for Cells
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352207
NACRES:
NA.75
Recommended Products
sterility
sterile-filtered
Quality Level
form
liquid
technique(s)
cell culture | mammalian: suitable
impurities
endotoxin, tested
components
L-glutamine: no
NaHCO3: no
HEPES: no
shipped in
dry ice
storage temp.
−20°C
General description
Cell Freezing Medium-Glycerol 1× is designed to protect and preserve cells during frozen storage. It is recommended for the cryopreservation of cells.
Application
Cell Freezing Medium-Glycerol 1× has been used for cryopreservation of honey bee semen, dispersed blastomeres, and whole blastoderms.
Other Notes
Complete ready-to-use medium. Proprietary formulation containing Minimum Essential Medium, glycerol, calf serum and fetal bovine serum.
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Usage of green coconut water and different tissue culture media for in vitro honey bee semen storage (Apis mellifera; Hymenoptera: Apoidea)
Almeida R, et al.
Interciencia, 27, 317-321 (2002)
Cryopreservation of rainbow trout blastoderms
BaBIak I, et al.
Cybium, 32, 139-141 (2008)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service