Skip to Content
Merck
All Photos(3)

Key Documents

C5625

Sigma-Aldrich

Lithium carbamoylphosphate dibasic hydrate

≥85% (enzymatic)

Synonym(s):

CAP-Li2, Carbamoylphosphate dilithium salt, Carbamyl phosphate dilithium salt, Carbamyl phosphoric acid dilithium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2COOPO3Li2·xH2O
CAS Number:
Molecular Weight:
152.89 (anhydrous basis)
Beilstein:
6128395
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25

product name

Lithium carbamoylphosphate dibasic hydrate,

Assay

≥85% (enzymatic)

form

powder

quality

may contain ~25% free acid (determined by PNMR)

color

white

storage temp.

−20°C

SMILES string

[Li+].[Li+].NC(=O)OP([O-])([O-])=O

InChI

1S/CH4NO5P.2Li/c2-1(3)7-8(4,5)6;;/h(H2,2,3)(H2,4,5,6);;/q;2*+1/p-2

InChI key

TUHZSMCNIUXAIJ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Carbamoylphosphate is used to study processes involving the mammalian urea cycle and nitrogen metabolism. Carbamoylphosphate is used as a substrate to identify and characterize ornithine transcarbamylase(s).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service