Skip to Content
Merck
All Photos(1)

Key Documents

C4987

Sigma-Aldrich

Ceramide phosphorylethanolamine

≥98% (TLC)

Synonym(s):

1-(2-Aminoethly)-phosphoryl-ceramide, Cer-PE, N-Acyl-Sphingosylphosphorylethanolamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H77N2O6P
Molecular Weight:
689.00
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (TLC)

form

powder

solubility

chloroform/methanol (2:1): soluble

shipped in

wet ice

storage temp.

−20°C

General description

Ceramide phosphorylethanolamine is a phosphosphingolipid.[1] It maintains the structural framework of the plasma membrane, aids in the formation and functioning of plasma membrane lipid rafts, cell signaling and transduction events. Ceramide phosphorylethanolamine modulates various cellular processes, like growth, differentiation, apoptosis and angiogenesis.[2]

Biochem/physiol Actions

Ceramide phosphorylethanolamine is a sphingolipid with sphingoid base backbones.[2][1] It is found in the rumen protozoan Entodinium caudatum , in the pupae of the green bottle fly, Lucilia Caesar, in adult housefly, Musca domestica and in few Mesogastropodoid snails.[1]
Sphingolipid with ethanolamine base, found in insect membranes.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Identification of ceramide phosphorylethanolamine and ceramide phosphorylglycerol in the lipids of an anaerobic bacterium
LaBach JP and White DC
Journal of Lipid Research, 10(5), 528-534 (1969)
Quantitation of ceramide phosphorylethanolamines containing saturated and unsaturated sphingoid base cores
Masood MA, et al.
Analytical Biochemistry, 400(2), 259-269 (2010)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service