Skip to Content
Merck
All Photos(3)

Documents

C4680

Sigma-Aldrich

α-Cyclodextrin

powder, BioReagent, suitable for cell culture, ≥98%

Synonym(s):

α-Schardinger dextrin, Cyclohexaamylose, Cyclomaltohexaose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H60O30
CAS Number:
Molecular Weight:
972.84
Beilstein:
79627
EC Number:
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.77

sterility

non-sterile

product line

BioReagent

Assay

≥98%

form

powder

technique(s)

cell culture | mammalian: suitable

mp

>278 °C (dec.) (lit.)

solubility

H2O: 10 mg/mL

shipped in

ambient

storage temp.

room temp

SMILES string

OC[C@H]1O[C@@H]2O[C@H]3[C@H](O)[C@@H](O)[C@H](O[C@@H]3CO)O[C@H]4[C@H](O)[C@@H](O)[C@H](O[C@@H]4CO)O[C@H]5[C@H](O)[C@@H](O)[C@H](O[C@@H]5CO)O[C@H]6[C@H](O)[C@@H](O)[C@H](O[C@@H]6CO)O[C@H]7[C@H](O)[C@@H](O)[C@H](O[C@@H]7CO)O[C@H]1[C@H](O)[C@H]2O

InChI

1S/C36H60O30/c37-1-7-25-13(43)19(49)31(55-7)62-26-8(2-38)57-33(21(51)15(26)45)64-28-10(4-40)59-35(23(53)17(28)47)66-30-12(6-42)60-36(24(54)18(30)48)65-29-11(5-41)58-34(22(52)16(29)46)63-27-9(3-39)56-32(61-25)20(50)14(27)44/h7-54H,1-6H2/t7-,8-,9-,10-,11-,12-,13-,14-,15-,16-,17-,18-,19-,20-,21-,22-,23-,24-,25-,26-,27-,28-,29-,30-,31-,32-,33-,34-,35-,36-/m1/s1

InChI key

HFHDHCJBZVLPGP-RWMJIURBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

α-Cyclodextrin (α-CD) is a cyclic oligosaccharide that consists of six glucopyranose units linked by α-(1,4) bonds. α-CD acts as a soluble dietary fiber and hence it is preferred to be widely used in food industry.

Application

Useful for the solubilization of nonpolar macromolecules such as fatty acids, lipids, and cholesterol for use in cell culture applications. α-Cyclodextrin has been used to solubilize odanacatib, a cathepsin K inhibitor.

Biochem/physiol Actions

α-Cyclodextrin is found to form a firm complex with dietary fats. This way it decreases the bioavailability and absorption of fats. It is known to regulate triglyceride and leptin levels in serum. In rat models, α-Cyclodextrin is shown to induce insulin sensitivity and fecal fat excretion. Thus, α-cyclodextrin is considered to be effective for treating obesity and metabolic syndromes.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pulsed low-dose RANKL as a potential therapeutic for postmenopausal osteoporosis
Cline SA, et al.
JCI insight, 1(13) (2016)
Functional Carbohydrates: Development, Characterization, and Biomanufacture (2017)
The Beneficial Effects a-Cyclodextrin on Blood Lipids and Weight Loss in Healthy Humans
Comerford KB, et al
Obesity (Silver Spring, Md.), 19(6), 1200-1204 (2011)
Zhongshu Li et al.
Chemical communications (Cambridge, England), 48(79), 9864-9866 (2012-08-07)
A novel deconvolution method for energy-resolved reaction cross sections is applied to determine intrinsic gas-phase dissociation energies for non-covalent α-cyclodextrin host-guest complexes. M06-2X//M06-L/6-31+G(d,p) calculations reproduce the experimental results and enable us to quantify the contribution of intermolecular hydrogen bonding.
Costanza Casati et al.
Journal of the American Chemical Society, 134(46), 19108-19117 (2012-10-31)
We present here the results of a CW-ESR investigation of a double spin labeled α-cyclodextrin-based [2]rotaxane that is characterized by the presence of nitroxide labels both at the wheel and at the dumbbell. This was accomplished by synthesizing a spin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service