C-39267
Smooth Muscle Cell Growth Medium 2
SupplementMix containing all media supplements pre-mixed in one vial, 1 Mix for 500 ml
Synonym(s):
Recommended with C-22262
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
form
aqueous solution
packaging
pkg of 1 vial
technique(s)
cell culture | mammalian: suitable
shipped in
dry ice
storage temp.
−20°C
General description
Smooth Muscle Cell Growth Medium 2 SupplementMix (basal medium sold separately)
To form a complete medium, it should be combined with the basal medium: C-22262
To form a complete medium, it should be combined with the basal medium: C-22262
Application
Smooth Muscle Cell Growth Medium 2 is a low-serum (5% V/V) medium developed to establish and maintain smooth muscle cell cultures from blood vessels and hollow organs. The medium is optimized for primary human cells but can also be used for bovine, porcine, murine, dog, and rat smooth muscle cells.Recommended for:Human Umbilical Artery Smooth Muscle Cells (HUASMC)Human Coronary Artery Smooth Muscle Cells (HCASMC)Human Pulmonary Artery Smooth Muscle Cells (HPASMC)Human Aortic Smooth Muscle Cells (HAoSMC)Human Bronchial Smooth Muscle Cells (HBSMC)Human Tracheal Smooth Muscle Cells (HTSMC)Human Uterine Smooth Muscle Cells (HUtSMC)
Components
Part of a Cell Culture System offered in a variety of formats:Growth Medium (Ready-to-use): Includes Basal Medium and SupplementMixGrowth Medium Kit: Includes Basal Medium and SupplementPackSupplementMix: Contains all media supplements premixed in one vialSupplementPack: Contains all media supplements as individual vials
Quality
All lots of PromoCell Smooth Muscle Cell Growth Medium are subjected to comprehensive quality control tests using primary human smooth muscle cells. Each lot is checked for growth promoting activity, adherence rate, and typical morphology of the tested smooth muscle cells. Approved in-house lots of media are used as a reference. In addition, all lots of media have been tested for the absence of microbial contaminants (fungi, bacteria, mycoplasma).
Preparation Note
Final supplement concentrations after addition to the basal medium:Fetal Calf Serum: 0.05 ml / mlEpidermal Growth Factor (recombinant human): 0.5 ng / mlBasic Fibroblast Growth Factor (recombinant human): 2 ng / mlInsulin (recombinant human): 5 µg / ml
Storage and Stability
Store the SupplementMix at -20°C in the dark immediately after arrival. If stored properly, it is stable until the expiry date stated on the label. After adding the supplements to the Basal Medium, the shelf life of the complete medium is 6 weeks at 4 to 8°C. Do not freeze the complete medium. For use, pre-warm only an aliquot of the complete medium and keep the remaining medium refrigerated at 4 to 8°C.
Subculture Routine
Click here for more information.
recommended
Product No.
Description
Pricing
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service