C-39016
Keratinocyte Growth Medium 2
SupplementMix containing all media supplements pre-mixed in one vial, 1 Mix for 500 ml
Synonym(s):
Recommended with C-20211
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352207
NACRES:
NA.71
Recommended Products
form
liquid
packaging
pkg of 1 vial
technique(s)
cell culture | mammalian: suitable
shipped in
dry ice
storage temp.
−20°C
General description
Keratinocyte Growth Medium 2 SupplementMix (basal medium sold separately)
To form a complete medium, it should be combined with the basal medium: C-20211
To form a complete medium, it should be combined with the basal medium: C-20211
Application
Keratinocyte Growth Medium 2 is a serum-free medium developed for the cultivation of epidermal keratinocytes without feeder cells. The medium is optimized for primary human cells but can also be used for horse and porcine keratinocytes, as well as mouse and rat keratinocytes after adapting the CaCl2 concentration. In addition, it is suited for rat corneal keratinocytes.Recommended for:Normal Human Epidermal Keratinocytes (NHEK), juvenile foreskinNormal Human Epidermal Keratinocytes (NHEK), adult donor
Components
Part of a Cell Culture System offered in a variety of formats:Growth Medium (Ready-to-use): Includes Basal Medium and SupplementMixGrowth Medium Kit: Includes Basal Medium and SupplementPackSupplementMix: Contains all media supplements premixed in one vialSupplementPack: Contains all media supplements as individual vials
Quality
All lots of PromoCell Keratinocyte Growth Medium 2 are subjected to comprehensive quality control tests using primary human keratinocytes. Each lot is checked for growth promoting activity, adherence rate, and typical morphology of the tested keratinocytes. Approved inhouse lots of media are used as a reference. In addition, all lots of media have been tested for the absence of microbial contaminants (fungi, bacteria, mycoplasma).
Preparation Note
Final supplement concentrations after addition to the basal medium:Bovine Pituitary Extract: 0.004 ml / mlEpidermal Growth Factor (recombinant human): 0.125 ng / mlInsulin (recombinant human): 5 µg / mlHydrocortisone: 0.33 µg / mlEpinephrine: 0.39 µg / mlTransferrin (recombinant human): 10 µg / mlCaCl2: 0.06 mMNote: CaCl2 is provided as separate stock solution. This allows users to adjust the CaCl2 concentration individually. To ensure the maximal growth performance of normal human Keratinocytes, PromoCell recommends adding the entire 60 μlCaCl2 stock solution, which results in final 0.06 mM CaCl2 concentration. In case a higher CaCl2 concentration is required (e.g. differentiation experiments), a 0.5 M CaCl2-Solution can be ordered separately (C-34006).
Storage and Stability
Store the SupplementMix at -20°C in the dark immediately after arrival. If stored properly, it is stable until the expiry date stated on the label. After adding the supplements to the Basal Medium, the shelf life of the complete medium is 6 weeks at 4 to 8°C. Do not freeze the complete medium. For use, pre-warm only an aliquot of the complete medium and keep the remaining medium refrigerated at 4 to 8°C.
Subculture Routine
Click here for more information.
recommended
Product No.
Description
Pricing
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service