Skip to Content
Merck
All Photos(4)

Key Documents

C3649

Sigma-Aldrich

CHAPSO

≥98%

Synonym(s):

3-([3-Cholamidopropyl]dimethylammonio)-2-hydroxy-1-propanesulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C32H58N2O8S
CAS Number:
Molecular Weight:
630.88
Beilstein:
5842642
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:
NACRES:
NA.25

description

zwitterionic

Assay

≥98%

form

powder

mol wt

micellar avg mol wt 7000

aggregation number

11

technique(s)

protein quantification: suitable

conductivity

≤100 μmho per cm at 24 °C (0.5 M aqueous solution)

CMC

8 mM (20-25°C)

mp

184-186 °C (lit.)

transition temp

cloud point 90 °C

SMILES string

[H][C@@]12C[C@H](O)CC[C@]1(C)[C@@]3([H])C[C@H](O)[C@]4(C)[C@]([H])(CC[C@@]4([H])[C@]3([H])[C@H](O)C2)[C@H](C)CCC(=O)NCCC[N+](C)(C)CC(O)CS([O-])(=O)=O

InChI

1S/C32H58N2O8S/c1-20(7-10-29(39)33-13-6-14-34(4,5)18-23(36)19-43(40,41)42)24-8-9-25-30-26(17-28(38)32(24,25)3)31(2)12-11-22(35)15-21(31)16-27(30)37/h20-28,30,35-38H,6-19H2,1-5H3,(H-,33,39,40,41,42)/t20-,21+,22-,23?,24-,25+,26+,27-,28+,30+,31+,32-/m1/s1

InChI key

GUQQBLRVXOUDTN-XOHPMCGNSA-N

Looking for similar products? Visit Product Comparison Guide

General description

CHAPSO is a non-denaturing zwitterionic bile salt derivative, derived from Chaps by the addition of a functional hydroxyl group.

Application

A nondenaturing zwitterionic detergent with characteristics similar to CHAPS, although it is more soluble due to a more polar head group. Useful for the solubilization of integral membrane proteins.
CHAPSO has been used:
  • in a study to assess the processes of liposome solubilization and reconstitution of two transport proteins
  • in a study to investigate a novel bicelle system for orienting macromolecules over a wide temperature range
  • as a mixture with dilauroyl phosphatidylcholine (DLPC) in solid-state nuclear magnetic resonance (NMR) technique to study membrane-associated proteins

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A liquid crystalline medium for measuring residual dipolar couplings over a wide range of temperatures.
Wang H, et al.
Journal of Biomolecular Nmr, 12(3), 443-446 (1998)
Magnetically orientable phospholipid bilayers containing small amounts of a bile salt analogue, CHAPSO.
Sanders CR and Prestegard J
Biophysical Journal, 58(2), 447-460 (1990)
S Cavagnero et al.
Journal of biomolecular NMR, 13(4), 387-391 (1999-06-03)
We have prepared and characterized a novel bicelle system composed of 1,2-di-O-dodecyl-sn-glycero-3-phosphocholine (DIODPC) and 3-(chloramidopropyl)dimethylammonio-2-hydroxyl-1-propane sulfonate (CHAPSO). At the optimal DIODPC/CHAPSO molar ratio of 4.3:1, this medium becomes magnetically oriented from pH 6.5 down to pH 1.0. Unlike previously reported
J Cladera et al.
European journal of biochemistry, 243(3), 798-804 (1997-02-01)
The process of liposome solubilization and reconstitution of two transport proteins have been studied using Chaps and Chapso (3-[(3-cholamidopropyl)dimethylammonio]-2- hydroxy-1-propanesulfonate). The solubilization of unilamellar liposomes was followed by absorption experiments and the process was shown to fit well to the
Satoko Osawa et al.
The Journal of biological chemistry, 283(28), 19283-19292 (2008-05-16)
gamma-Secretase is an aspartic protease that hydrolyzes type I membrane proteins within the hydrophobic environment of the lipid bilayer. Using the CHAPSO-solubilized gamma-secretase assay system, we previously found that gamma-secretase activity was sensitive to the concentrations of detergent and phosphatidylcholine.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service