Skip to Content
Merck
All Photos(1)

Key Documents

C3535

Sigma-Aldrich

Complement C8 from human serum

suitable for radioiodination, RIA, ≥85% (SDS-PAGE), ≥125,000 C8H50 units/mg protein (using C8 deficient serum)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.61

biological source

human

Quality Level

Assay

≥85% (SDS-PAGE)

form

solution

specific activity

≥125,000 C8H50 units/mg protein (using C8 deficient serum)

concentration

1 mg/mL in PBS, pH 7.2

technique(s)

immunoelectrophoresis: suitable

suitability

suitable for radioiodination, RIA

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... C8A(731)

Application

Complement C8 is one of the end terminals of the complement system contained in the membrane attack complex (MAC). In the (MAC), C8 initiates and coordinates MAC pore formation. It has been used in research to investigate the mechanisms of how C8-C9 and C9-C9 interactions facilitate folding of the membrane attack complex perforin (MACPF) domain to form the circular pore.

Biochem/physiol Actions

C8 is one of the components of the terminal complement complex and is required for a functional complement system. It is composed of disulfide-linked C8α and C8γ and a noncovalently associated C8β chain. During formation of the membrane-attack complex, a domain on C8α induces C9 to polymerize and form a pore-like structure.

Quality

Functionally pure by a sensitive hemolytic assay using depleted sera.

Other Notes

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel J Slade et al.
Biochemistry, 45(16), 5290-5296 (2006-04-19)
Human C8 is one of five components of the membrane attack complex of complement (MAC). It contains three subunits (C8alpha, C8beta, C8gamma) arranged as a disulfide-linked C8alpha-gamma dimer that is noncovalently associated with C8beta. C8alpha, C8beta, and complement components C6
Doryen Bubeck et al.
Journal of molecular biology, 405(2), 325-330 (2010-11-16)
Complement component C8 plays a pivotal role in the formation of the membrane attack complex (MAC), an important antibacterial immune effector. C8 initiates membrane penetration and coordinates MAC pore formation. High-resolution structures of C8 subunits have provided some insight into
Leslie L Lovelace et al.
The Journal of biological chemistry, 286(20), 17585-17592 (2011-04-02)
C8 is one of five complement proteins that assemble on bacterial membranes to form the lethal pore-like "membrane attack complex" (MAC) of complement. The MAC consists of one C5b, C6, C7, and C8 and 12-18 molecules of C9. C8 is
Zhexu Chi et al.
Molecular cell, 80(1), 43-58 (2020-09-17)
Immune cell function depends on specific metabolic programs dictated by mitochondria, including nutrient oxidation, macromolecule synthesis, and post-translational modifications. Mitochondrial adaptations have been linked to acute and chronic inflammation, but the metabolic cues and precise mechanisms remain unclear. Here we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service