Skip to Content
Merck
All Photos(1)

Key Documents

C3411

Sigma-Aldrich

2′,7′-Bis(2-carboxyethyl)-5(6)-carboxyfluorescein

≥90% (HPLC)

Synonym(s):

BCECF

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H20O11
CAS Number:
Molecular Weight:
520.44
Beilstein:
8375770
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.32

Quality Level

Assay

≥90% (HPLC)

form

powder

storage temp.

−20°C

SMILES string

OC(=O)CCc1cc2c(OC3=CC(=O)C(CCC(O)=O)=CC3=C2c4cc(ccc4C(O)=O)C(O)=O)cc1O

InChI

1S/C27H20O11/c28-19-10-21-17(7-12(19)2-5-23(30)31)25(16-9-14(26(34)35)1-4-15(16)27(36)37)18-8-13(3-6-24(32)33)20(29)11-22(18)38-21/h1,4,7-11,28H,2-3,5-6H2,(H,30,31)(H,32,33)(H,34,35)(H,36,37)

InChI key

DKROGDOAOXDINY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2′,7′-Bis(2-carboxyethyl)-5(6)-carboxyfluorescein has been used in a fluorescence-based screening assay, to determine the pH.

Biochem/physiol Actions

2′,7′-Bis(2-carboxyethyl)-5(6)-carboxyfluorescein (BCECF) is useful to study multidrug resistance-associated protein (MRP) transporter action within the cell. BCECF is known to interfere with drug efflux interactions.
Fluorescent indicator for measurement of cytoplasmic pH in lymphocytes.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alexander A Tokmakov et al.
Biochemical and biophysical research communications, 517(1), 140-145 (2019-07-20)
Mitogen-activated protein kinases (MAPKs) are involved in the regulation of various cellular processes, including cell survival and apoptosis. Here, we report that Xenopus p42 MAPK becomes phosphorylated in apoptotic eggs, however this modification does not activate the enzyme. Using phosphorylation
Adelaide S M Dennis et al.
Scientific reports, 8(1), 8795-8795 (2018-06-13)
Four hundred structurally diverse drug-like compounds comprising the Medicines for Malaria Venture's 'Pathogen Box' were screened for their effect on a range of physiological parameters in asexual blood-stage malaria (Plasmodium falciparum) parasites. Eleven of these compounds were found to perturb
High-throughput workflow for monitoring and mining bioprocess data: Application to infer E. coli physiological response to perturbations.
Heux S, et al.
Applied and Environmental Microbiology, AEM-05838 (2011)
Patrick Hofmann et al.
Cardiovascular research, 115(1), 230-242 (2018-08-15)
Long non-coding RNAs (lncRNAs) have been shown to regulate numerous processes in the human genome, but the function of these transcripts in vascular aging is largely unknown. We aim to characterize the expression of lncRNAs in endothelial aging and analyse
Chang-Yin Lee et al.
Oncology reports, 37(1), 399-407 (2016-11-15)
Panax notoginseng (P. notoginseng) and its components are used as traditional Chinese medicine for cardiovascular disease, although studies concerning the anti-metastatic properties of these compounds are limited. The goal of this study was to investigate the effects of notoginsenoside R1 (NGR1), an important

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service