Skip to Content
Merck
All Photos(2)

Key Documents

C271

Sigma-Aldrich

CX546

≥98% (HPLC), solid

Synonym(s):

1-(1,4-Benzodioxan-6-ylcarbonyl)piperidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H17NO3
CAS Number:
Molecular Weight:
247.29
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

storage condition

protect from light

color

white to off-white

solubility

DMSO: ≥10 mg/mL

storage temp.

2-8°C

SMILES string

O=C(N1CCCCC1)c2ccc3OCCOc3c2

InChI

1S/C14H17NO3/c16-14(15-6-2-1-3-7-15)11-4-5-12-13(10-11)18-9-8-17-12/h4-5,10H,1-3,6-9H2

InChI key

LJUNPHMOGNFFOS-UHFFFAOYSA-N

Gene Information

General description

CX546, a benzoylpiperidine derivative ampakine is an analog of CX516.

Application

CX546 has been used as positive allosteric modulator for the glutamatergic receptor α-amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid (AMPA) in mice.

Biochem/physiol Actions

CX546 has antipsychotic functionality and has the potential to treat schizophrenia. It improves the defects associated with the prepulse inhibition (PPI) and latent inhibition (LI) in mice lacking metabotropic glutamate receptor type 5 (mGluR5). Additionally, CX546 potentiates synaptic plasticity, elicits neuroprotection and promotes the neurotrophin expression.
Positive AMPA glutamate receptor modulator.

Caution

Photosensitive

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hiroaki Sacai et al.
Nature communications, 11(1), 5140-5140 (2020-10-14)
Autism spectrum disorder (ASD) is thought to result from deviation from normal development of neural circuits and synaptic function. Many genes with mutation in ASD patients have been identified. Here we report that two molecules associated with ASD susceptibility, contactin
Laureen D Hachem et al.
Stem cells and development, 26(23), 1675-1681 (2017-09-28)
Transplantation of neural stem/progenitor cells (NSPCs) following spinal cord injury (SCI) is a promising strategy to enhance regeneration but is limited by poor survival of grafted cells. Determining methods to enhance survival of NSPCs is therefore essential. Positive modulation of
Amy C Arai et al.
The Journal of pharmacology and experimental therapeutics, 303(3), 1075-1085 (2002-11-20)
CX516 (BDP-12) and CX546, two first-generation benzamide-type AMPA receptor modulators, were compared with regard to their influence on AMPA receptor-mediated currents, autaptic responses in cultured hippocampal neurons, hippocampal excitatory postsynaptic currents, synaptic field potentials, and agonist binding. The two drugs
C Lu et al.
Molecular psychiatry, 21(2), 159-168 (2015-11-26)
Numerous risk genes have recently been implicated in susceptibility to autism and schizophrenia. Translating such genetic findings into disease-relevant neurobiological mechanisms is challenging due to the lack of throughput assays that can be used to assess their functions on an
The ampakine CX546 restores the prepulse inhibition and latent inhibition deficits in mGluR5-deficient mice
Lipina T, et al.
Neuropsychopharmacology, 32(4), 745-756 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service