C2249
Human Collagen Type I
from human fibroblasts, liquid, 3 mg/mL, suitable for cell culture
Synonym(s):
Collagen Solution, Fibroblast Collagen
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352202
NACRES:
NA.75
Recommended Products
Product Name
Collagen Solution from human fibroblasts, 3 mg/mL, sterile-filtered, BioReagent, suitable for cell culture
biological source
human fibroblasts
Quality Level
sterility
sterile-filtered
product line
BioReagent
form
solution
packaging
pkg of 20 mL
concentration
3 mg/mL
technique(s)
cell culture | mammalian: suitable
surface coverage
6‑10 μg/cm2
Binding Specificity
Peptide Source: Collagen
Peptide Source: Elastin
foreign activity
endotoxin ≤5.0 μmole/min-mg protein
shipped in
wet ice
storage temp.
2-8°C
General description
Collagen constitutes to one-third of the total protein in humans. Collagen fibril is the basic component of the tissues containing collagen. It is predominantly expressed in the extracellular matrix.Type I collagen is a major structural component of skin, bone, tendon, and other fibrous connective tissues. It differs from other collagens by its low lysine hydroxylation and low carbohydrate composition. Type I collagen is a heterotrimer composed of two α1(I) chains and one alpha2(I) chain, which spontaneously form a triple helix scaffold at neutral pH and 37 °C. Purified human collagen solution is produced from neo-natal human fibroblasts that have secreted human extracellular matrix (hECM) using a propriety cell culture production system. This product contains a high monomer content. In vitro cultures were prepared using intensively tested human fibroblast cells and purified using a manufacturing process following applicable aspects of cGMP. The human fibroblast cells have undergone significant testing demonstrating safety from viruses and other adventitious agents. This process contains built-in, validated steps to ensure the inactivation of possible prion and/or viral contaminants.
Application
Collagen Solution from human fibroblasts has been used
- as a component of coating solution for human bone marrow chip fabrication.
- in fibroblast-like synoviocytes culturing.
- as a coating solution for microchannels for organ chip culture
Biochem/physiol Actions
Collagen is an essential ingredient of connective tissue. Studies in a Chinese family show that mutation in COL1A1 (collagenase type I) is linked with type I osteogenesis imperfecta. Collagen is linked with subchondral turnover of bone, and might have potential as marker to determine the state of joint space narrowing and osteophytes in osteoarthritis.
Proteins belonging to the collagen family supports and nourishes body tissues such as tendons, cartilage, skin and sclera of the eye. Collagen type 1 is a versatile building material required for tissue elasticity and maintains its stability and strength. Collagen is also present in byssus thread of invertebrates, which helps in surface attachment. Defective collagen affects the biomechanical property of arterial tissues.
Storage Class Code
10 - Combustible liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Aspek Klinik, Genetik dan Molekuler Osteogensis Imperfekta
Mariska M and Auerkari EI
Journal of Dentistry Indonesia, 14(2), 95-110 (2015)
Collagen in arterial walls: biomechanical aspects
Collagen, 285-324 (2008)
Synovitis biomarkers: ex vivo characterization of three biomarkers for identification of inflammatory osteoarthritis
Kjelgaard PC, et al.
Biomarkers, 20(8), 547-556 (2015)
On-chip recapitulation of clinical bone-marrow toxicities and patient-specific pathophysiology
Chou DB, et al.
Nature Biomedical Engineering, 4(4), 394?406-394?406 (2020)
Frederick Grinnell
Trends in cell biology, 13(5), 264-269 (2003-05-14)
Research on fibroblast biology in three-dimensional collagen matrices offers new opportunities to understand the reciprocal and adaptive interactions that occur between cells and surrounding matrix in a tissue-like environment. Such interactions are integral to the regulation of connective tissue morphogenesis
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service