Skip to Content
Merck
All Photos(2)

Key Documents

C117

Sigma-Aldrich

5-Carboxamidotryptamine maleate salt

powder, ≥98% (HPLC)

Synonym(s):

5-CT, AH-21467

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H13N3O · C4H4O4
CAS Number:
Molecular Weight:
319.31
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to off-white

solubility

H2O: 10 mg/mL

storage temp.

−20°C

SMILES string

OC(=O)\C=C/C(O)=O.NCCc1c[nH]c2ccc(cc12)C(N)=O

InChI

1S/C11H13N3O.C4H4O4/c12-4-3-8-6-14-10-2-1-7(11(13)15)5-9(8)10;5-3(6)1-2-4(7)8/h1-2,5-6,14H,3-4,12H2,(H2,13,15);1-2H,(H,5,6)(H,7,8)/b;2-1-

InChI key

PZQZSWAOVAMBQM-BTJKTKAUSA-N

Biochem/physiol Actions

Agonist at 5-HT1A, 5-HT1B, 5-HT1D, 5-HT5 and 5-HT7 serotonin receptors.

comparable product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daria Guseva et al.
Inflammatory bowel diseases, 20(9), 1516-1529 (2014-07-30)
Intestinal inflammation is often associated with an increased level of serotonin (5-HT), an important gastrointestinal signaling molecule involved in gut homeostasis through stimulation of specific receptors. In this study, we investigated the role of 5-HT7 receptor (5-HT7R) in the induction
N J MacDonald et al.
British journal of clinical pharmacology, 30(1), 107-113 (1990-07-01)
1. Zabicipril, S9650, a new angiotensin converting enzyme inhibitor, was administered to salt-replete, normotensive males in single doses of up to 10 mg. 2. The safety, tolerance and dose-response relationship with regard to inhibition of plasma ACE activity were characterised
Hao Zhang et al.
International immunopharmacology, 52, 93-100 (2017-09-11)
Chronic inflammation is a major contributing factor in the pathogenesis of many diseases. Natural product berberine (BBR) exhibits potent anti-inflammatory effect in vitro and in vivo, while the underlying mechanisms remain elusive. Sirt1, a NAD
Ayako Moritomo et al.
Bioorganic & medicinal chemistry, 22(15), 4323-4337 (2014-06-12)
We previously reported that the novel dual 5-HT₂B and 5-HT7 receptor antagonist N-(9-hydroxy-9H-fluorene-2-carbonyl)guanidine (4) exerted a suppressing effect on 5-HT-induced dural protein extravasation in guinea pigs. To develop a synthetic strategy, we performed docking studies of lead compound 4 bound
W L Bacon et al.
The Journal of pharmacology and experimental therapeutics, 294(2), 672-679 (2000-07-20)
The 5-hydroxytryptamine(7) (5-HT(7)) receptor was originally defined by molecular biology techniques. The 5-HT(7) receptor protein and mRNA are found in brain areas, such as the CA3 subfield of the hippocampus, that are involved in various neuropsychiatric disease states. No functional

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service