Skip to Content
Merck
All Photos(1)

Key Documents

C0992

Sigma-Aldrich

Anti-Cy3/Cy5 antibody, Mouse monoclonal

clone CY-96, purified from hybridoma cell culture

Synonym(s):

Monoclonal Anti-Cy3/Cy5, Cy3 Antibody, Cy3 Antibody - Anti-Cy3/Cy5 antibody, Mouse monoclonal

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

CY-96, monoclonal

form

buffered aqueous solution

concentration

~1.5 mg/mL

technique(s)

direct ELISA: suitable
dot blot: 1-2 μg/mL using cell protein extracts labeded with Cy3 or Cy5
immunocytochemistry: suitable
immunoprecipitation (IP): suitable
microarray: suitable

isotype

IgG2a

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

Anti-Cy3/Cy5 antibody, Mouse monoclonal, (mouse IgG2a isotype) is derived from the hybridoma CY-96 produced by the fusion of mouse myeloma cells (NS1 cells) and splenocytes from Balb/c mice immunized with a mixture of proteins labeled with Cy3 or Cy5.

Specificity

The antibody recognizes Cy3 and Cy5 conjugated to proteins.

Immunogen

mixture of proteins labeled with Cy3/Cy5.

Application

Anti-Cy3/Cy5 antibody, Mouse monoclonal has been used in:
  • immunofluorescence
  • western blot
  • dot blot
  • enzyme linked immunosorbent assay (ELISA)
  • immunoprecipitation
  • immunocytochemistry
  • protein microarrays
  • In in situ hybridization

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunofluorescence (1 paper)

Biochem/physiol Actions

Cy 3 and Cy5 are the most popular cyanine dyes, used combined for two color detection. Cy3 dyes are fluorescent orange while Cy5 is fluorescent in the red region. Cyanine belonging to polymethine group. CyDyes are a family of fluorophores that can be used for labeling proteins, peptides, DNA, RNA, and other biomolecules. These dyes are small, pH insensitive, soluble in aqueous solution and are tolerant to DMSO. They are more photostable than fluorescein, have high molar extinction coefficients and favorable quantum yields. Mainly Cy3 and Cy5 are used in many different biological assays such as DNA microarrays, protein microarrays, two-dimensional protein analysis (2D gels), fluorescence resonance energy transfer (FRET), and immunocytochemistry.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Enders
Acta neurochirurgica. Supplement, 89, 9-13 (2004-09-01)
Microarray analysis has been emerged as a tool to characterize the overall reaction of cells in culture or tissue to different stimuli e.g. stressful events by analysing bulk RNA present at a particular time point. It has supplemented or even
An Improved Method for Cell Type-Selective Glycomic Analysis of Tissue Sections Assisted by Fluorescence Laser Microdissection
Nagai-Okatani C, et al.
International Journal of Molecular Sciences, 20(3), 700-700 (2019)
Chiaki Nagai-Okatani et al.
International journal of molecular sciences, 20(3) (2019-02-10)
Lectin microarray (LMA) is a highly sensitive technology used to obtain the global glycomic profiles of endogenous glycoproteins in biological samples including formalin-fixed paraffin-embedded tissue sections. Here, we describe an effective method for cell type-selective glycomic profiling of tissue fragments
Transplantation of ovarian granulosa-like cells derived from human induced pluripotent stem cells for the treatment of murine premature ovarian failure
Liu T. et al.
Molecular Medicine Reports, 13(6), 5053-5058 (2016)
A K Kenworthy
Methods (San Diego, Calif.), 24(3), 289-296 (2001-06-14)
Fluorescence resonance energy transfer (FRET) detects the proximity of fluorescently labeled molecules over distances >100 A. When performed in a fluorescence microscope, FRET can be used to map protein-protein interactions in vivo. We here describe a FRET microscopy method that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service