Skip to Content
Merck
All Photos(2)

Documents

B9904

Sigma-Aldrich

Anti-Mouse IgG (Fc specific)–Biotin antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

conjugate

biotin conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse

concentration

≥2 mg/mL

technique(s)

direct ELISA: 1:50,000
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:300
western blot: 1:200,000-1:400,000 using total cell extract of HeLa cells

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

IgG is present in large quantities in the human serum. It constitutes about 10-20% of the plasma proteins. IgG is composed of glycoproteins, out of which it is 82-96% proteins and 4-18% carbohydrates. It consists of four sub-classes i.e IgG1, IgG2, IgG3, and IgG4. IgG is composed of four polypeptide chains-two heavy chains (γchains) and two light chains (κ or λ chains) which are linked by inter-chain disulfide bonds. The heavy chains consist of a N-terminal variable domain (VH) and three constant domains (CH1, CH2, CH3). A hinge region exists between the CH1 and CH2 region. The light chains have one N-terminal variable domain (VL) and one constant domain (CL). The heavy and the light chains are linked at VH and CH1 domain to form the Fab arm (Fragment antigen binding). The antigen binds to the V regions of the antibody.

Application

Anti-Mouse IgG (Fc specific)-Biotin antibody has been used in enzyme linked immunosorbent assay (ELISA), dot blot, immunohistology, nuclear labeling, ELISA-inhibition method. immunohistochemistry.
Anti-Mouse IgG (Fc specific)-Biotin antibody produced in goat was used for immunocytochemistry staining to visualize neonatal myosin in chicken muscle sections. It was used for ELISPOT analysis to study IgG levels in mice infected with cholera toxin.

Biochem/physiol Actions

IgG antibody subtype is the most abundant of serum immunoglobulins of the immune system. It is secreted by B cells and is found in blood and extracellular fluids and provides protection from infections caused by bacteria, fungi and viruses. Maternal IgG is transferred to fetus through the placenta that is vital for immune defense of the neonate against infections. The coupling of biotin to Anti-Mouse IgG (Fc specific) antibody allows for the binding of various labels such as avidin or streptavidin.

Other Notes

Antibody adsorbed with human IgG and rat serum proteins.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Preparation Note

Adsorbed to reduce background staining with human or rat samples.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kate E Broderick et al.
Human vaccines, 7 Suppl, 22-28 (2011-01-26)
Electropermeabilization of mammalian cells is a technique that has been used for the delivery of therapeutics, such as DNA plasmids or DNA vaccines. Typically, delivery via electropermeabilization occurs through injection of the substance into the tissue of interest followed by
Malika Guendouz et al.
Pharmaceutical biology, 55(1), 2145-2152 (2017-10-07)
Royal jelly (RJ) has long been used to promote human health. The current study investigated the preventive effects of RJ against the development of a systemic and intestinal immune response in mice allergic to cow's milk proteins. Balb/c mice treated
A TgCRND8 mouse model of Alzheimer?s disease exhibits sexual dimorphisms in behavioral indices of cognitive reserve
Granger MW, et al.
Journal of Alzheimer'S Disease, 51(3), 757-773 (2016)
IgE Cross Reactivity Between Reindeer and Bovine Milk ss-Lactoglobulins in Cow's Milk Allergic Patients
Suutari T J, et al.
Journal of investigative medicine high impact case reports, 16(5), 296-296 (2006)
André Steinecke et al.
Development (Cambridge, England), 141(2), 460-471 (2014-01-02)
Inhibitory interneurons control the flow of information and synchronization in the cerebral cortex at the circuit level. During embryonic development, multiple subtypes of cortical interneurons are generated in different regions of the ventral telencephalon, such as the medial and caudal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service