B9638
Basal Medium Eagle
With Earle′s salts and L-glutamine, without sodium bicarbonate, powder, suitable for cell culture
Synonym(s):
BME
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352207
NACRES:
NA.75
Recommended Products
form
powder
technique(s)
cell culture | mammalian: suitable
components
phenol red: 0.011 g/L
NaHCO3: no
L-glutamine: 0.292 g/L
Earle’s salts (5% CO2): yes
shipped in
ambient
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
Application
BME, when properly supplemented, has demonstrated wide applicability, for supporting monolayer growth of a wide variety of normal and transformed cell lines.
Quantity
Formulated to contain 9.2 grams of powder per liter of medium.
Reconstitution
Supplement with 2.2 g/L sodium bicarbonate.
Other Notes
Eagle's Basal Medium or BME, developed by Harry Eagle, is one of the most widely used of all synthetic cell culture media.
also commonly purchased with this product
Product No.
Description
Pricing
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Alkaline phosphatase downregulation promotes lung adenocarcinoma metastasis via the c-Myc/RhoA axis.
Zhefeng Lou et al.
Cancer cell international, 21(1), 217-217 (2021-04-17)
Lung adenocarcinoma (LUAD) metastasis significantly reduces patient survival; hence inhibiting the metastatic ability of lung cancer cells will greatly prolong patient survival. Alkaline phosphatase (ALPL), a homodimeric cell surface phosphohydrolase, is reported to play a controversial role in prostate cancer
Qian Wu et al.
PloS one, 10(5), e0127703-e0127703 (2015-05-23)
Tumor necrosis factor (TNF)-α is a cytokine involved in acute inflammatory phase reactions, and is the primary upstream mediator in the cochlear inflammatory response. Treatment of the organ of Corti with TNF-α can induce hair cell damage. However, the resulting
Gurudutt Pendyala et al.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 42(7), 1435-1446 (2017-01-20)
Emerging epidemiology studies indicate that maternal immune activation (MIA) resulting from inflammatory stimuli such as viral or bacterial infections during pregnancy serves as a risk factor for multiple neurodevelopmental disorders including autism spectrum disorders and schizophrenia. Although alterations in the
M K Nanjappa et al.
Andrology, 5(6), 1165-1173 (2017-10-27)
Stem Leydig cells (SLCs), precursors of testicular Leydig cells that secrete testosterone required for male sexual differentiation, spermatogenesis, and fertility, were recently identified in rat testes. Various types of stem cells have shown the ability to differentiate into other tissues
Lizhen Wang et al.
Neurobiology of aging, 29(3), 357-367 (2006-12-13)
beta-Site APP cleavage enzyme 1 (BACE1) is the beta-secretase responsible for generating amyloid-beta (A beta) peptides in Alzheimer's disease (AD). Previous studies suggest that activation of protein kinase C (PKC) modulates the beta-secretase-mediated cleavage of APP and reduces the production
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service