B7652
Betamethasone 21-phosphate disodium
≥97%
Synonym(s):
Betamethasone disodium phosphate, Betamethasone sodium phosphate
About This Item
Recommended Products
sterility
non-sterile
Quality Level
Assay
≥97%
form
powder
solubility
water: 50 mg/mL, clear to very slightly hazy, faintly yellow
shipped in
ambient
storage temp.
room temp
SMILES string
[Na+].[Na+].[H][C@@]12CCC3=CC(=O)C=C[C@]3(C)[C@@]1(F)[C@@H](O)C[C@@]4(C)[C@@]2([H])C[C@H](C)[C@]4(O)C(=O)COP([O-])([O-])=O
InChI
1S/C22H30FO8P.2Na/c1-12-8-16-15-5-4-13-9-14(24)6-7-19(13,2)21(15,23)17(25)10-20(16,3)22(12,27)18(26)11-31-32(28,29)30;;/h6-7,9,12,15-17,25,27H,4-5,8,10-11H2,1-3H3,(H2,28,29,30);;/q;2*+1/p-2/t12-,15-,16-,17-,19-,20-,21-,22-;;/m0../s1
InChI key
PLCQGRYPOISRTQ-LWCNAHDDSA-L
Gene Information
human ... NR3C1(2908)
Looking for similar products? Visit Product Comparison Guide
Biochem/physiol Actions
Diagnostic Implications
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Repr. 1B - STOT RE 2
Target Organs
Liver,Kidney,Endocrine system
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service