Skip to Content
Merck
All Photos(3)

Key Documents

B0274

Sigma-Aldrich

5-Bromo-4-chloro-3-indolyl phosphate p-toluidine salt

tablet

Synonym(s):

BCIP® p-toluidine salt, X-phosphate p-toluidine salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H6BrClNO4P · C7H9N
CAS Number:
Molecular Weight:
433.62
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83

form

tablet

storage temp.

−20°C

SMILES string

Cc1ccc(N)cc1.OP(O)(=O)Oc2c[nH]c3ccc(Br)c(Cl)c23

InChI

1S/C8H6BrClNO4P.C7H9N/c9-4-1-2-5-7(8(4)10)6(3-11-5)15-16(12,13)14;1-6-2-4-7(8)5-3-6/h1-3,11H,(H2,12,13,14);2-5H,8H2,1H3

InChI key

QEIFSLUFHRCVQL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Specificity

Substrate of choice for use with alkaline phosphatase in immunoblotting and, less commonly, in immunohistological staining procedures. High assay sensitivity is achieved via amplification when BCIP is used in conjunction with Nitro Blue Tetrazolium (NBT) Tablets (Catalog No. N5514). This substrate produces an insoluble end product that is blue-purple in color and can be observed visually.

Application

For the colorimetric detection of alkaline phosphatase-labeled molecules, 5-Bromo-4-chloro-3-indolyl phosphate (BCIP) and Nitro Blue Tetrazolium (NBT) are available. The BCIP/NBT substrate system is versatile and functions in a variety of applications, including Northern Southern, and Western blotting, in situ hybridization, and immunohistochemistry.
BCIP is provided in two salt forms: the disodium salt which is soluble in water and the p-toluidine form which is soluble in dimethylformamide. These salt forms may be used to prepare a stock solution that in combination with NBT and a reaction buffer, form a substrate solution for alkaline phosphatase. This substrate system, when incubated with alkaline phosphatase, produces an insoluble NBT diformazan product that is easily observable with its purple color. For added convenience, a mixture of BCIP and NBT is provided in an easily dissolvable tablet form or as a ready-to-use liquid.
For the colorimetric detection of alkaline phosphatase-labeled molecules.

Quantity

Contains 25 mg substrate per tablet.

Substrates

Histochemical substrate for alkaline phosphatase.

Legal Information

BCIP is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Carolyn Byrne et al.
Methods in molecular biology (Clifton, N.J.), 289, 127-136 (2004-10-27)
The skin as a surface organ is uniquely accessible for whole embryo/fetal analyzes of developmental changes, such as gene induction, protein expression, formation of epidermal-derived appendages such as hair follicles, and formation of the protective barrier. Such analyses have emphasized
Audrey Fouchs et al.
Biology of the cell, 102(8), 447-456 (2010-04-10)
Activation of MAPKs (mitogen-activated protein kinases), in particular ERK1/2 (extracellular-signal-regulated kinase 1/2), has been reported to take place in a large variety of cell types after hypo-osmotic cell swelling. Depending on cell type, ERK1/2 phosphorylation can then serve or not
Substrates for cytochemical demonstration of enzyme activity. II. Some dihalo-3-indolyl phosphates and sulfates.
J P Horwitz et al.
Journal of medicinal chemistry, 9(3), 447-447 (1966-05-01)
Michael Alifrangis et al.
Malaria journal, 3, 16-16 (2004-06-15)
The aim of this study was to develop site-specific antibodies as a tool to capture Plasmodium falciparum-dihydrofolate reductase (Pf-DHFR) from blood samples from P. falciparum infected individuals in order to detect, in a sandwich ELISA, structural alterations due to point
M S Blake et al.
Analytical biochemistry, 136(1), 175-179 (1984-01-01)
A rapid, sensitive method has been developed to detect antibody-antigen complexes on "Western blots." The methods of H. Towbin, T. Staehlin, and J. Gordon were used to separate and blot the antigens onto nitrocellulose. The remaining sites of attachment were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service