AXYEV200R
Pipette tips for Tecan® EVO/ Caliper Zephyr® Automated Liquid Handling System
racked, capacity 200 μL, pkg of 5x960tips/cs, clear
Synonym(s):
pipet tips
About This Item
Recommended Products
manufacturer/tradename
Axygen Inc EV-200-R
Corning
packaging
pkg of 5x960tips/cs
capacity
200 μL
color
clear
Looking for similar products? Visit Product Comparison Guide
Related Categories
Other Notes
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service