Skip to Content
Merck
All Photos(1)

Key Documents

AV54329

Sigma-Aldrich

Anti-IL4 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-BSF1, Anti-IL-4, Anti-Interleukin 4, Anti-MGC79402

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

15 kDa

species reactivity

human

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... IL4(3565)

Immunogen

Synthetic peptide directed towards the middle region of human IL4

Application

Anti-IL4 antibody produced in rabbit is suitable for western blotting at a concentration of 1μg/mL.

Biochem/physiol Actions

Interleukin-4 (IL-4) is a T cell derived chemokine that stimulates the Th2 mediated immune response and proliferation of B cells. The effects of IL-4 are mediated by two types of receptors, type I receptor consisting of IL-4Rα and common γ chain and type II receptor composed of IL-4Rα and IL-13Rα1. The signalling stimulated by IL-4 leads to activation of JAK/STAT6 and IRS-mediated PI3K/Akt pathway. Through these pathways, IL-4 is responsible for endocytic activity of macrophages, chemotaxis of leukocytes in response to inflammation, angiogenesis and regulation of nitric oxide metabolism in macrophages. Anti-tumor effects of IL-4 have been reported in cancers of breast, liver and renal cells.

Sequence

Synthetic peptide located within the following region: TVLRQFYSHHEKDTRCLGATAQQFHRHKQLIRFLKRLDRNLWGLAGLNSC

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michal Kuczma et al.
Journal of immunotoxicology, 11(4), 319-327 (2013-12-20)
Immunotherapy is becoming an increasingly attractive therapeutic alternative for conventional cancer therapy. In recent years Foxp3(+) regulatory T-cells (T(R)) were identified as the major obstacle to effective cancer immunotherapy. The abundance of these cells in peripheral blood is increased in
C K Oh et al.
European respiratory review : an official journal of the European Respiratory Society, 19(115), 46-54 (2010-10-20)
Asthma is a complex, persistent, inflammatory disease characterised by airway hyperresponsiveness in association with airway inflammation. Studies suggest that regular use of high-dose inhaled corticosteroids and long-acting bronchodilators or omalizumab (a humanised monoclonal antibody that binds to immunoglobulin E and
Kerstin Göbel et al.
European journal of immunology, 44(8), 2295-2305 (2014-05-09)
Lymphocyte adhesion and subsequent trafficking across endothelial barriers are essential steps in various immune-mediated disorders of the CNS, including MS. The molecular mechanisms underlying these processes, however, are still unknown. Phospholipase D1 (PLD1), an enzyme that generates phosphatidic acid through
Boo-Young Kim et al.
The Laryngoscope, 124(10), E399-E404 (2014-04-15)
In this study, we addressed the immunotherapeutic potential of human placental extract (HPE) in a murine allergic rhinitis (AR) model and explored its immunological mechanisms. In vivo study using an animal model. HPE was administered to BALB/c mice before sensitization
Agnes Hamzaoui et al.
Immunobiology, 219(11), 873-879 (2014-08-17)
Vitamin D [25(OH)D3] deficiency has been associated with asthma as in many inflammatory and autoimmune pathologies; however, there is still a lack of data about the effects of administration of vitamin D in immune regulation in young asthmatic patients. In

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service