Skip to Content
Merck
All Photos(2)

Documents

AV46626

Sigma-Aldrich

Anti-RTN2 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-NSP2, Anti-NSPL1, Anti-Reticulon 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

51 kDa

species reactivity

human, rat, pig, horse, mouse, bovine, dog

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RTN2(6253)

Immunogen

Synthetic peptide directed towards the N terminal region of human RTN2

Application

Anti-RTN2 antibody produced in rabbit is suitable for western blotting at a concentration of 2.5μg/mL. It is also useful for immunohistochemistry at a concentration of 4-8μg/mL.

Biochem/physiol Actions

RTN2 (reticulon 2) gene also referred to as NSP2 or NSPL1 is a member of reticulon encoding gene family. It plays a pivotal role in organizing endoplasmic reticulum and distal motor axons. RTN2B, isoform of RTN2 regulates the trafficking as well as facilitates as a positive modulator for delivering the EAAC1 (excitatory amino acid carrier 1) from ER to the cell surface. Mutation in RTN2 gene leads to axon-degenerative disorder hereditary spastic paraplegia type 12.

Sequence

Synthetic peptide located within the following region: MGQVLPVFAHCKEAPSTASSTPDSTEGGNDDSDFRELHTAREFSEEDEEE

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Niamh C O'Sullivan et al.
Human molecular genetics, 21(15), 3356-3365 (2012-05-01)
Several causative genes for hereditary spastic paraplegia encode proteins with intramembrane hairpin loops that contribute to the curvature of the endoplasmic reticulum (ER), but the relevance of this function to axonal degeneration is not understood. One of these genes is
Gladys Montenegro et al.
The Journal of clinical investigation, 122(2), 538-544 (2012-01-11)
Hereditary spastic paraplegias (HSPs) are a group of genetically heterogeneous neurodegenerative conditions. They are characterized by progressive spastic paralysis of the legs as a result of selective, length-dependent degeneration of the axons of the corticospinal tract. Mutations in 3 genes
Yiting Liu et al.
The Journal of biological chemistry, 283(10), 6561-6571 (2007-12-22)
Excitatory amino acid transporters (EAATs) are the primary regulators of extracellular glutamate concentrations in the central nervous system. Their dysfunction may contribute to several neurological diseases. To date, five distinct mammalian glutamate transporters have been cloned. In brain, EAAC1 (excitatory

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service