Skip to Content
Merck
All Photos(2)

Documents

AV41675

Sigma-Aldrich

Anti-CYP2D6 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-CPD6, Anti-CYP2D, Anti-CYP2D@, Anti-CYP2DL1, Anti-Cytochrome P450, family 2, subfamily D, polypeptide 6, Anti-MGC120389, Anti-MGC120390, Anti-RP4-669P10.2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

55 kDa

species reactivity

human, rat

concentration

0.5 mg - 1 mg/mL

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CYP2D6(1565)

Related Categories

General description

Anti-CγP2D6 polyclonal antibody reacts with pig, rabbit, human, mouse, rat, and zebrafish cytochrome P450, family 2, subfamily D, polypeptide 6 proteins.
Cytochrome P450, family 2, subfamily D, polypeptide 6 (CγP2D6) is an important oxidizing enzyme involved in drug metabolism wherein it sometimes functions as a drug activator. CγP2D6 metabolizes drugs in important classes including antidepressants (tricyclics, tetracyclics, SSRIs, SNRIs); opioids; antipsychotics; β-blockers; class I antiarrhythmics and a variety of other drug applications.

Immunogen

Synthetic peptide directed towards the N terminal region of human CYP2D6

Application

Anti-CγP2D6 polyclonal antibody is used to tag cytochrome P450, family 2, subfamily D, polypeptide 6 proteins for detection and quantitation by Western blotting and in cells and tissues by immunohistochemical (IHC) techniques. It is used as a probe to study the role of cytochrome P450, family 2, subfamily D, polypeptide 6 proteins in metabolism of many important therapeutic drugs.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunofluorescence (1 paper)

Biochem/physiol Actions

CYP2D6 is a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein localizes to the endoplasmic reticulum and is known to metabolize as many as 20% of commonly prescribed drugs. Its substrates include debrisoquine, an adrenergic-blocking drug; sparteine and propafenone, both anti-arrythmic drugs; and amitryptiline, an anti-depressant.This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein localizes to the endoplasmic reticulum and is known to metabolize as many as 20% of commonly prescribed drugs. Its substrates include debrisoquine, an adrenergic-blocking drug; sparteine and propafenone, both anti-arrythmic drugs; and amitryptiline, an anti-depressant. The gene is highly polymorphic in the population; certain alleles result in the poor metabolizer phenotype, characterized by a decreased ability to metabolize the enzyme′s substrates. The gene is located near two cytochrome P450 pseudogenes on chromosome 22q13.1. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.

Sequence

Synthetic peptide located within the following region: RPPVPITQILGFGPRSQGVFLARYGPAWREQRRFSVSTLRNLGLGKKSLE

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nobuhito Mori et al.
Scientific reports, 10(1), 5646-5646 (2020-04-15)
Although various production methods for 3D vascularised tissues have been developed, constructing capillary-like structures branching from perfusable large channels remains difficult. This study describes a method to fabricate tube-shaped 3D liver-like tissue (tubular liver tissue) with large channels and capillary-like
Arnaud Carpentier et al.
Stem cell research, 16(3), 640-650 (2016-04-12)
The establishment of protocols to differentiate human pluripotent stem cells (hPSCs) including embryonic (ESC) and induced pluripotent (iPSC) stem cells into functional hepatocyte-like cells (HLCs) creates new opportunities to study liver metabolism, genetic diseases and infection of hepatotropic viruses (hepatitis
Chang Li et al.
Nature communications, 14(1), 1613-1613 (2023-03-25)
Dopamine has a significant role in motor and cognitive function. The dopaminergic pathways originating from the midbrain have received the most attention; however, the relevance of the cerebellar dopaminergic system is largely undiscovered. Here, we show that the major cerebellar
Saiyong Zhu et al.
Nature, 508(7494), 93-97 (2014-02-28)
Human induced pluripotent stem cells (iPSCs) have the capability of revolutionizing research and therapy of liver diseases by providing a source of hepatocytes for autologous cell therapy and disease modelling. However, despite progress in advancing the differentiation of iPSCs into

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service