Key Documents
ADF356302L-EP
Adapter for Eppendorf® F-35-6-30 Rotor
holds 1 x 2.6 - 7 mL round-bottom tube and blood collection tubes, large rotor bore, large rotor bore, pack of 2 ea
About This Item
UNSPSC Code:
41103911
feature
holds 1 x 2.6 - 7 mL round-bottom tube and blood collection tubes, large rotor bore
packaging
pack of 2 ea
manufacturer/tradename
Eppendorf® 5427733000 (EU)
technique(s)
centrifugation: suitable
compatibility
for use with Rotor F-35-6-30 (large rotor bore)
Related Categories
General description
Adapters to fit various size tubes and bottles for use with Eppendorf rotor F-35-6-30.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)