Skip to Content
Merck
All Photos(1)

Key Documents

AAPMLS

Sigma-Aldrich

Amino Acid Peptide Metabolite Library Of Standards

Supplied by IROA Technologies

Synonym(s):

AAPMLS, Mass Spectrometry Metabolite Library

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.21

Quality Level

packaging

pkg of 2 plates

manufacturer/tradename

IROA Technologies

plate size

96 wells

shipped in

dry ice

storage temp.

−20°C

General description

Compounds can either be used as standards and injected individually or a strategy may be employed to pool compounds to allow multiple mixture to be analyzed per injection.

Application

AAPMLS (Amino Acid Peptide Metabolite Library of Standards) is a collection of high quality small biochemical molecules in an economical, ready-to-use format. This library of standards is most commonly used for metabolite qualification and quantification for MS.

Features and Benefits

168 unique small molecule organic acid metabolites covering key metabolic pathways including the following classes of compounds:
  • Acetylated, methylated and hydroxyl amino acids
  • Di Peptides

To view a complete list of compounds in the library for recent lots, download the plate maps below:

The complete plate map with Name, parent CID, molecular formula, molecular weight, CAS, ChEBI, HMDB ID, PubChem Compound and Substance ID are provided with the software download.

Preparation Note

Compounds are pre-weighed and provided at 5 μg per well, enough for multiple injections. Arranged in 96-well polypropylene plates with alphanumerically assigned positions. Discovery software is provided to help construct libraries, useful for metabolomic compound ID.

Other Notes

For additional information on our range of Biochemicals, please complete this form.
MLSDiscovery software tool is provided to support the extraction, manipulation, and storage of the data generated when using the AAPMLS and other authentic libraries of metabolite standards supplied by IROA Technologies. Software comes with user manual and training videos.

To download the software package, click here - MLSDiscovery software.

To view the MLSDiscovery Video visit: MLSDiscovery Tutorial Video.

Legal Information

AAPMLS is a trademark of IROA Technologies LLC
MLSDiscovery is a trademark of IROA Technologies LLC

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service