Skip to Content
Merck
All Photos(4)

Key Documents

A9795

Sigma-Aldrich

Adenine hydrochloride hydrate

powder, BioReagent, suitable for cell culture

Synonym(s):

6-Aminopurine hydrochloride hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H5N5 · HCl · xH2O
CAS Number:
Molecular Weight:
171.59 (anhydrous basis)
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.75

biological source

synthetic (organic)

product line

BioReagent

Assay

≥99%

form

powder

technique(s)

cell culture | mammalian: suitable

solubility

H2O: 50 mg/mL

SMILES string

O.Cl.Nc1ncnc2[nH]cnc12

InChI

1S/C5H5N5.ClH.H2O/c6-4-3-5(9-1-7-3)10-2-8-4;;/h1-2H,(H3,6,7,8,9,10);1H;1H2

InChI key

MYRDTAUFFBYTHA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Adenine hydrochloride hydrate has been used:
  • As a component of FAD cell culture media
  • In maintaining organotypic cultures for recreating stratified epithelium from primary cell cultures
  • In limbal-corneal epithelial culturing and keratinocytes

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vitronectin supports migratory responses of corneal epithelial cells to substrate bound IGF-I and HGF, and facilitates serum-free cultivation
Ainscough SL, et al.
Experimental Eye Research, 83(6), 1505-1514 (2006)
Functional investigations of keratinocyte stem cells and progenitors at a single-cell level using multiparallel clonal microcultures
Epidem. Inf., 83(6), 13-23 (2010)
Three-dimensional organotypic culture of stratified epithelia
Rhim Andrew D and Rustgi Anil K
Cold Spring Harbor Protocols, 2015(4) (2015)
Felipe Perozzo Daltoe et al.
Brazilian oral research, 34, e033-e033 (2020-04-09)
The aim of our study was to isolate populations of keratinocyte stem cells based on the expression of cell surface markers and to investigate whether the culture could affect their phenotype. keratinocytes from human oral mucosa were sorted based on
Elham Nili et al.
Molecules (Basel, Switzerland), 26(22) (2021-11-28)
A recombinant formulation of silk fibroin containing the arginine-glycine-aspartic acid (RGD) cell-binding motif (RGD-fibroin) offers potential advantages for the cultivation of corneal cells. Thus, we investigated the growth of corneal stromal cells and epithelial cells on surfaces created from RGD-fibroin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service